Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590633_m1
          See other SLC5A6 GE Assays ›
          Gene Symbol:
          Slc5a6
          Gene Name:
          solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
          Gene Aliases:
          SMVT
          Chromosome Location:
          Chr.6: 26685823 - 26697110 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590633_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc5a6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130746.1 NP_570102.1 13 - 14 1688 61
          GenBank mRNA AF026554.1 - 13 - 14 1688 61
          AF081204.2 - 10 - 11 1276 61
          AF081205.1 - 10 - 11 1276 61
          BC168145.1 - 11 - 12 1550 8371761 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000295.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc5a6

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          biotin transport
          pantothenate transmembrane transport
          sodium-dependent multivitamin transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1053947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122510 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1053947 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1053947 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1053947 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700504192 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31761 - -
          Agilent Rat cDNA Microarray (G4105A) 700504192 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10883188 - -
          Affymetrix Rat Gene 1.0 ST Array 10883192 - -
          Affymetrix Rat Gene 1.0 ST Array 10883181 - -
          Affymetrix Rat Gene 1.0 ST Array 10883184 - -
          Affymetrix Rat Gene 1.0 ST Array 10883189 - -
          Affymetrix Rat Gene 1.0 ST Array 10883178 - -
          Affymetrix Rat Gene 1.0 ST Array 10883193 - -
          Affymetrix Rat Gene 1.0 ST Array 10883194 - -
          Affymetrix Rat Gene 1.0 ST Array 10883176 - -
          Affymetrix Rat Gene 1.0 ST Array 10883187 - -
          Affymetrix Rat Gene 1.0 ST Array 10883190 - -
          Affymetrix Rat Gene 1.0 ST Array 10883177 - -
          Affymetrix Rat Gene 1.0 ST Array 10883182 - -
          Affymetrix Rat Gene 1.0 ST Array 10883191 - -
          Affymetrix Rat Gene 1.0 ST Array 10883183 - -
          Affymetrix Rat Gene 1.0 ST Array 10883186 - -
          Affymetrix Rat Gene 1.0 ST Array 10883180 - -
          Affymetrix Rat Gene 1.0 ST Array 10883179 - -
          Affymetrix Rat Gene 1.0 ST Array 10883195 - -
          Affymetrix Rat Gene 1.0 ST Array 10883185 - -
          Affymetrix Rat Gene 1.1 ST Array 10883175 - -
          Rat Genome 230 2.0 Array 1367815_at - -
          Rat Genome 230 2.0 Array 1367815_PM_at - -
          Affymetrix HT Rat Focus Array 1367815_at - -
          Affymetrix HT Rat Focus Array 1367815_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600001938.rn.1 TC0600000295.rn.1 RTA TC
          Rat Expression Set 230 1367815_at - -
          Rat Expression Set 230 1367815_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10883175 - -
          Rat Genome U34 Set AF081204_s_at - -
          Rat Genome U34 Set AF026554_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21879886 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358092 - -
          RatRef-12 Expression BeadChip ILMN_55398 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline