Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590969_m1
          See other CAV2 GE Assays ›
          Gene Symbol:
          Cav2
          Gene Name:
          caveolin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 44573264 - 44580640 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590969_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cav2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131914.2 NP_571989.2 2 - 3 455 72
          GenBank mRNA AF439780.1 - 2 - 3 332 72
          BC062059.1 - 2 - 3 455 5598269 72
          FQ209775.1 - 2 - 3 509 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000426.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cav2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          vesicle fusion
          mitochondrion organization
          endoplasmic reticulum organization
          regulation of mitotic nuclear division
          chemical synaptic transmission
          negative regulation of cell proliferation
          insulin receptor signaling pathway
          vesicle organization
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          vesicle docking
          skeletal muscle fiber development
          positive regulation of peptidyl-tyrosine phosphorylation
          protein oligomerization
          positive regulation of dopamine receptor signaling pathway
          caveola assembly
          positive regulation of protein localization to nucleus
          SNARE binding
          protein binding
          protein kinase binding
          syntaxin binding
          protein binding, bridging
          mitogen-activated protein kinase kinase kinase binding
          D1 dopamine receptor binding
          protein complex scaffold
          protein homodimerization activity
          protein heterodimerization activity
          phosphoprotein binding
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152825 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152825 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P441434 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853814 - -
          Affymetrix Rat Gene 1.0 ST Array 10931797 - -
          Affymetrix Rat Gene 1.0 ST Array 10853812 - -
          Affymetrix Rat Gene 1.0 ST Array 10853813 - -
          Affymetrix Rat Gene 1.0 ST Array 10931796 - -
          Affymetrix Rat Gene 1.0 ST Array 10853811 - -
          Affymetrix Rat Gene 1.0 ST Array 10853815 - -
          Affymetrix Rat Gene 1.0 ST Array 10931794 - -
          Affymetrix Rat Gene 1.0 ST Array 10931798 - -
          Affymetrix Rat Gene 1.0 ST Array 10931795 - -
          Affymetrix Rat Gene 1.1 ST Array 10931793 - -
          Affymetrix Rat Gene 1.1 ST Array 10853810 - -
          Rat Genome 230 2.0 Array 1377642_PM_at - -
          Rat Genome 230 2.0 Array 1386261_PM_x_at - -
          Rat Genome 230 2.0 Array 1370135_PM_at - -
          Rat Genome 230 2.0 Array 1390800_a_at - -
          Rat Genome 230 2.0 Array 1390800_PM_a_at - -
          Rat Genome 230 2.0 Array 1370135_at - -
          Rat Genome 230 2.0 Array 1377642_at - -
          Rat Genome 230 2.0 Array 1386261_x_at - -
          Affymetrix HT Rat Focus Array 1377642_PM_at - -
          Affymetrix HT Rat Focus Array 1377642_at - -
          Affymetrix HT Rat Focus Array 1370135_at - -
          Affymetrix HT Rat Focus Array 1370135_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400003018.rn.1 TC0400000426.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003021.rn.1 TC0400000426.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003020.rn.1 TC0400000426.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003022.rn.1 TC0400000426.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003023.rn.1 TC0400000426.rn.1 RTA TC
          Rat Expression Set 230 1370135_at - -
          Rat Expression Set 230 1370135_PM_at - -
          Rat Expression Set 230 1386261_x_at - -
          Rat Expression Set 230 1390800_a_at - -
          Rat Expression Set 230 1377642_at - -
          Rat Expression Set 230 1377642_PM_at - -
          Rat Expression Set 230 1390800_PM_a_at - -
          Rat Expression Set 230 1386261_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931793 - -
          Affymetrix Rat Gene 1.0 ST Array 10853810 - -
          Rat Genome U34 Set rc_AI010103_at - -
          Rat Genome U34 Set rc_AI101672_at - -
          Rat Genome U34 Set rc_AA818495_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21281328 - -
          ABI Rat Genome Survey Microarray 22296519 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374468 - -
          RatRef-12 Expression BeadChip ILMN_67467 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline