Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591028_m1
          See other HEMGN GE Assays ›
          Gene Symbol:
          Hemgn
          Gene Name:
          hemogen
          Gene Aliases:
          Edag-1, Edag1, Hgn, RP59
          Chromosome Location:
          Chr.5: 62004271 - 62025072 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591028_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hemgn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133294.1 NP_579828.1 1 - 2 164 95
          XM_008763693.1 XP_008761915.1 1 - 2 240 95
          XM_008763694.1 XP_008761916.1 2 - 3 308 95
          GenBank mRNA AJ302650.2 - 1 - 2 164 95
          BC078739.1 - 2 - 3 264 7111835 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002685.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hemgn

          Gene Ontology Categories:

          Function(s)

          multicellular organism development
          cell differentiation
          regulation of osteoblast differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412096 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19078 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054886 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054886 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054886 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P450846 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701279220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84635 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13629 - -
          Agilent Rat cDNA Microarray (G4105A) 701279220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876588 - -
          Affymetrix Rat Gene 1.0 ST Array 10876593 - -
          Affymetrix Rat Gene 1.0 ST Array 10876590 - -
          Affymetrix Rat Gene 1.0 ST Array 10876592 - -
          Affymetrix Rat Gene 1.0 ST Array 10876587 - -
          Affymetrix Rat Gene 1.0 ST Array 10876589 - -
          Affymetrix Rat Gene 1.0 ST Array 10876591 - -
          Affymetrix Rat Gene 1.1 ST Array 10876586 - -
          Rat Genome 230 2.0 Array 1387756_s_at - -
          Rat Genome 230 2.0 Array 1387756_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387756_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387756_s_at - -
          Affymetrix Rat Clariom S Assay PSR0500019989.rn.1 TC0500002685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019987.rn.1 TC0500002685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019988.rn.1 TC0500002685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019990.rn.1 TC0500002685.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019993.rn.1 TC0500002685.rn.1 RTA TC
          Rat Expression Set 230 1387756_s_at - -
          Rat Expression Set 230 1387756_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10876586 - -
          Rat Genome U34 Set rc_AI229683_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22261562 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357584 - -
          RatRef-12 Expression BeadChip ILMN_59672 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline