Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591060_m1
          See other GPNMB GE Assays ›
          Gene Symbol:
          Gpnmb
          Gene Name:
          glycoprotein (transmembrane) nmb
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 78694447 - 78715685 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591060_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpnmb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133298.1 NP_579832.1 9 - 10 1583 67
          GenBank mRNA AF184983.2 - 9 - 10 1583 67
          BC061725.1 - 9 - 10 1526 5598392 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000875.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpnmb

          Panther Classification:

          Molecular Function -

          membrane-bound signaling molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          cell adhesion
          bone mineralization
          integrin binding
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P990376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65462 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120360 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P990376 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P990376 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P990376 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P990376 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700631669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91484 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70092 - -
          Agilent Rat cDNA Microarray (G4105A) 700631669 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855455 - -
          Affymetrix Rat Gene 1.0 ST Array 10855451 - -
          Affymetrix Rat Gene 1.0 ST Array 10855450 - -
          Affymetrix Rat Gene 1.0 ST Array 10855461 - -
          Affymetrix Rat Gene 1.0 ST Array 10855456 - -
          Affymetrix Rat Gene 1.0 ST Array 10855457 - -
          Affymetrix Rat Gene 1.0 ST Array 10855460 - -
          Affymetrix Rat Gene 1.0 ST Array 10855458 - -
          Affymetrix Rat Gene 1.0 ST Array 10855453 - -
          Affymetrix Rat Gene 1.0 ST Array 10855452 - -
          Affymetrix Rat Gene 1.0 ST Array 10855459 - -
          Affymetrix Rat Gene 1.0 ST Array 10855454 - -
          Affymetrix Rat Gene 1.1 ST Array 10855449 - -
          Rat Genome 230 2.0 Array 1368187_PM_at - -
          Rat Genome 230 2.0 Array 1368187_at - -
          Affymetrix HT Rat Focus Array 1368187_PM_at - -
          Affymetrix HT Rat Focus Array 1368187_at - -
          Affymetrix Rat Clariom S Assay PSR0400006279.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006280.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006283.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006281.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006285.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006277.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006282.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006284.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006274.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006275.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006276.rn.1 TC0400000875.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006278.rn.1 TC0400000875.rn.1 RTA TC
          Rat Expression Set 230 1368187_at - -
          Rat Expression Set 230 1368187_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855449 - -
          Rat Genome U34 Set rc_AA997841_at - -
          Rat Genome U34 Set rc_AA851814_at - -
          Rat Genome U34 Set rc_AI112012_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22105497 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370437 - -
          RatRef-12 Expression BeadChip ILMN_67552 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline