Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591164_m1
          See other PRDM4 GE Assays ›
          Gene Symbol:
          Prdm4
          Gene Name:
          PR domain containing 4
          Gene Aliases:
          SC-1
          Chromosome Location:
          Chr.7: 23959929 - 23986535 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591164_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133312.1 NP_579846.1 1 - 2 133 66
          XM_006241160.2 XP_006241222.1 3 - 4 595 66
          XM_006241161.1 XP_006241223.1 3 - 4 594 66
          XM_008765213.1 XP_008763435.1 3 - 4 600 66
          GenBank mRNA AF176023.1 - 1 - 2 133 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000384.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdm4

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of cell growth
          cellular response to nutrient
          methylation
          cellular response to growth factor stimulus
          DNA binding
          death receptor binding
          methyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P397292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168936 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151385 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P397292 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P397292 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P397292 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17611 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11849 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894521 - -
          Affymetrix Rat Gene 1.0 ST Array 10894513 - -
          Affymetrix Rat Gene 1.0 ST Array 10894519 - -
          Affymetrix Rat Gene 1.0 ST Array 10894514 - -
          Affymetrix Rat Gene 1.0 ST Array 10894517 - -
          Affymetrix Rat Gene 1.0 ST Array 10894522 - -
          Affymetrix Rat Gene 1.0 ST Array 10894516 - -
          Affymetrix Rat Gene 1.0 ST Array 10894518 - -
          Affymetrix Rat Gene 1.0 ST Array 10894520 - -
          Affymetrix Rat Gene 1.0 ST Array 10894515 - -
          Affymetrix Rat Gene 1.1 ST Array 10894512 - -
          Rat Genome 230 2.0 Array 1369236_PM_at - -
          Rat Genome 230 2.0 Array 1369236_at - -
          Rat Genome 230 2.0 Array 1374736_at - -
          Rat Genome 230 2.0 Array 1374736_PM_at - -
          Affymetrix HT Rat Focus Array 1369236_at - -
          Affymetrix HT Rat Focus Array 1369236_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700003289.rn.1 TC0700000384.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700003290.rn.1 TC0700000384.rn.1 RTA TC
          Rat Expression Set 230 1369236_at - -
          Rat Expression Set 230 1369236_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894512 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21180113 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367684 - -
          RatRef-12 Expression BeadChip ILMN_64767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline