Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591259_m1
          See other ZFP111 GE Assays ›
          Gene Symbol:
          Zfp111
          Gene Name:
          zinc finger protein 111
          Gene Aliases:
          rKr2
          Chromosome Location:
          Chr.1: 81035686 - 81047083 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591259_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp111
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133323.1 NP_579857.1 3 - 4 463 108
          XM_006228375.2 XP_006228437.1 1 - 2 403 108
          XM_006228376.2 XP_006228438.1 2 - 3 559 108
          GenBank mRNA FQ211918.1 - 3 - 4 529 108
          U27186.1 - 3 - 4 463 108

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp111

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035041 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035041 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035041 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P252507 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P635860 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10914 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719640 - -
          Affymetrix Rat Gene 1.0 ST Array 10719644 - -
          Affymetrix Rat Gene 1.0 ST Array 10719638 - -
          Affymetrix Rat Gene 1.0 ST Array 10719635 - -
          Affymetrix Rat Gene 1.0 ST Array 10719636 - -
          Affymetrix Rat Gene 1.0 ST Array 10719643 - -
          Affymetrix Rat Gene 1.0 ST Array 10719637 - -
          Affymetrix Rat Gene 1.0 ST Array 10719642 - -
          Affymetrix Rat Gene 1.0 ST Array 10719639 - -
          Affymetrix Rat Gene 1.0 ST Array 10719634 - -
          Affymetrix Rat Gene 1.0 ST Array 10719641 - -
          Affymetrix Rat Gene 1.0 ST Array 10719645 - -
          Affymetrix Rat Gene 1.1 ST Array 10719633 - -
          Rat Genome 230 2.0 Array 1369052_PM_at - -
          Rat Genome 230 2.0 Array 1369052_at - -
          Affymetrix HT Rat Focus Array 1369052_at - -
          Affymetrix HT Rat Focus Array 1369052_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100039196.rn.1 TC0100004949.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100039197.rn.1 TC0100004949.rn.1 RTA TC
          Rat Expression Set 230 1369052_PM_at - -
          Rat Expression Set 230 1369052_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719633 - -
          Rat Genome U34 Set U27186_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21624224 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364784 - -
          RatRef-12 Expression BeadChip ILMN_52414 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline