Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591270_m1
          See other NAA35 GE Assays ›
          Gene Symbol:
          Naa35
          Gene Name:
          N(alpha)-acetyltransferase 35, NatC auxiliary subunit
          Gene Aliases:
          Mak10, T4a
          Chromosome Location:
          Chr.17: 5411479 - 5463898 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591270_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Naa35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133324.1 NP_579858.1 21 - 22 2102 66
          XM_006253526.2 XP_006253588.1 21 - 22 2367 66
          XM_006253527.1 XP_006253589.1 19 - 20 1966 66
          GenBank mRNA AF272892.1 - 21 - 22 2102 66
          BC074005.1 - 21 - 22 2079 7099269 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001229.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Naa35

          Panther Classification:

          Molecular Function -

          acetyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s)

          negative regulation of apoptotic process
          smooth muscle cell proliferation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P429781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104436 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P429781 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P429781 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P429781 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P429781 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700523847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91185 - -
          Agilent Rat cDNA Microarray (G4105A) 700523847 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797080 - -
          Affymetrix Rat Gene 1.0 ST Array 10797070 - -
          Affymetrix Rat Gene 1.0 ST Array 10797084 - -
          Affymetrix Rat Gene 1.0 ST Array 10797066 - -
          Affymetrix Rat Gene 1.0 ST Array 10797063 - -
          Affymetrix Rat Gene 1.0 ST Array 10797078 - -
          Affymetrix Rat Gene 1.0 ST Array 10797077 - -
          Affymetrix Rat Gene 1.0 ST Array 10797071 - -
          Affymetrix Rat Gene 1.0 ST Array 10797081 - -
          Affymetrix Rat Gene 1.0 ST Array 10797075 - -
          Affymetrix Rat Gene 1.0 ST Array 10797083 - -
          Affymetrix Rat Gene 1.0 ST Array 10797069 - -
          Affymetrix Rat Gene 1.0 ST Array 10797072 - -
          Affymetrix Rat Gene 1.0 ST Array 10797065 - -
          Affymetrix Rat Gene 1.0 ST Array 10797064 - -
          Affymetrix Rat Gene 1.0 ST Array 10797068 - -
          Affymetrix Rat Gene 1.0 ST Array 10797074 - -
          Affymetrix Rat Gene 1.0 ST Array 10797076 - -
          Affymetrix Rat Gene 1.0 ST Array 10797082 - -
          Affymetrix Rat Gene 1.0 ST Array 10797067 - -
          Affymetrix Rat Gene 1.0 ST Array 10797073 - -
          Affymetrix Rat Gene 1.0 ST Array 10797079 - -
          Affymetrix Rat Gene 1.0 ST Array 10797085 - -
          Affymetrix Rat Gene 1.1 ST Array 10797062 - -
          Rat Genome 230 2.0 Array 1398804_PM_at - -
          Rat Genome 230 2.0 Array 1398804_at - -
          Affymetrix HT Rat Focus Array 1398804_at - -
          Affymetrix HT Rat Focus Array 1398804_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700008495.rn.1 TC1700001229.rn.1 RTA TC
          Rat Expression Set 230 1398804_PM_at - -
          Rat Expression Set 230 1398804_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797062 - -
          Rat Genome U34 Set rc_AA851658_at - -
          Rat Genome U34 Set rc_AA851658_g_at - -
          Rat Genome U34 Set rc_AI180418_at - -
          Rat Genome U34 Set rc_AA819860_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20702342 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370276 - -
          RatRef-12 Expression BeadChip ILMN_69665 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline