Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591819_m1
          See other IL13RA2 GE Assays ›
          Gene Symbol:
          Il13ra2
          Gene Name:
          interleukin 13 receptor, alpha 2
          Gene Aliases:
          IL-13R-alpha-2
          Chromosome Location:
          Chr.X: 118443958 - 118514716 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591819_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il13ra2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133538.2 NP_598222.2 12 - 13 1604 137
          XM_006257427.2 XP_006257489.1 11 - 12 1472 137
          XM_006257428.2 XP_006257490.1 11 - 12 1590 137
          XM_006257430.2 XP_006257492.1 10 - 11 1403 137
          XM_006257431.2 XP_006257493.1 8 - 9 1111 137
          XM_008773388.1 XP_008771610.1 11 - 12 1744 137
          XM_008773389.1 XP_008771611.1 12 - 13 1631 137
          XM_008773390.1 XP_008771612.1 8 - 9 1205 137
          XM_008773391.1 XP_008771613.1 8 - 9 1812 137
          GenBank mRNA AF448818.1 - 12 - 13 1604 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002308.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il13ra2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of immunoglobulin production
          immunoglobulin mediated immune response
          cytokine-mediated signaling pathway
          negative regulation of mast cell degranulation
          cellular response to mechanical stimulus
          cytokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116043 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109028 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46168 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98946 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937293 - -
          Affymetrix Rat Gene 1.0 ST Array 10937281 - -
          Affymetrix Rat Gene 1.0 ST Array 10937290 - -
          Affymetrix Rat Gene 1.0 ST Array 10937282 - -
          Affymetrix Rat Gene 1.0 ST Array 10937291 - -
          Affymetrix Rat Gene 1.0 ST Array 10937280 - -
          Affymetrix Rat Gene 1.0 ST Array 10937283 - -
          Affymetrix Rat Gene 1.0 ST Array 10937287 - -
          Affymetrix Rat Gene 1.0 ST Array 10937295 - -
          Affymetrix Rat Gene 1.0 ST Array 10937301 - -
          Affymetrix Rat Gene 1.0 ST Array 10937284 - -
          Affymetrix Rat Gene 1.0 ST Array 10937298 - -
          Affymetrix Rat Gene 1.0 ST Array 10937299 - -
          Affymetrix Rat Gene 1.0 ST Array 10937288 - -
          Affymetrix Rat Gene 1.0 ST Array 10937285 - -
          Affymetrix Rat Gene 1.0 ST Array 10937300 - -
          Affymetrix Rat Gene 1.0 ST Array 10937297 - -
          Affymetrix Rat Gene 1.0 ST Array 10937296 - -
          Affymetrix Rat Gene 1.0 ST Array 10937289 - -
          Affymetrix Rat Gene 1.0 ST Array 10937286 - -
          Affymetrix Rat Gene 1.0 ST Array 10937294 - -
          Affymetrix Rat Gene 1.1 ST Array 10937292 - -
          Affymetrix Rat Gene 1.1 ST Array 10937279 - -
          Rat Genome 230 2.0 Array 1369266_at - -
          Rat Genome 230 2.0 Array 1369266_PM_at - -
          Affymetrix HT Rat Focus Array 1369266_at - -
          Affymetrix HT Rat Focus Array 1369266_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00014445.rn.1 TC0X00002308.rn.1 RTA TC
          Rat Expression Set 230 1369266_at - -
          Rat Expression Set 230 1369266_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937279 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21733855 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376826 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline