Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591943_m1
          See other MLXIPL GE Assays ›
          Gene Symbol:
          Mlxipl
          Gene Name:
          MLX interacting protein-like
          Gene Aliases:
          ChREBP, WS-bHLH, Wbscr14, bHLHd14
          Chromosome Location:
          Chr.12: 24590645 - 24619639 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591943_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mlxipl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133552.1 NP_598236.1 6 - 7 1114 84
          GenBank mRNA AB074517.1 - 6 - 7 1114 84
          FN432819.1 - 6 - 7 1081 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001358.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mlxipl

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of glycolytic process
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          response to glucose
          glucose mediated signaling pathway
          negative regulation of peptidyl-serine phosphorylation
          glucose homeostasis
          positive regulation of fatty acid biosynthetic process
          positive regulation of glycolytic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of lipid biosynthetic process
          fatty acid homeostasis
          negative regulation of cell cycle arrest
          cellular response to glucose stimulus
          negative regulation of oxidative phosphorylation
          regulation of energy homeostasis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein kinase binding
          carbohydrate response element binding
          protein homodimerization activity
          sequence-specific DNA binding
          protein heterodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P755887 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P755887 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P755887 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P755887 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P755887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940564 - -
          Affymetrix Rat Gene 1.1 ST Array 10940564 - -
          Rat Genome 230 2.0 Array 1368614_PM_at - -
          Rat Genome 230 2.0 Array 1368614_at - -
          Rat Genome 230 2.0 Array 1377013_PM_at - -
          Rat Genome 230 2.0 Array 1377013_at - -
          Affymetrix HT Rat Focus Array 1368614_at - -
          Affymetrix HT Rat Focus Array 1368614_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200011745.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011736.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011743.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011738.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011750.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011740.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011749.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011735.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011751.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011741.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011752.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011734.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011737.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011739.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011742.rn.1 TC1200001358.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200011748.rn.1 TC1200001358.rn.1 RTA TC
          Rat Expression Set 230 1368614_PM_at - -
          Rat Expression Set 230 1368614_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940564 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21687967 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline