Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591979_m1
          See other CSF2RB GE Assays ›
          Gene Symbol:
          Csf2rb
          Gene Name:
          colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage)
          Gene Aliases:
          Csf2rb1
          Chromosome Location:
          Chr.7: 119554383 - 119568248 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591979_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csf2rb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133555.1 NP_598239.1 7 - 8 1186 64
          GenBank mRNA AJ000555.1 - 7 - 8 1186 64
          S79263.1 - 7 - 8 1186 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001411.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csf2rb

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          response to lipopolysaccharide
          molecular_function
          cytokine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398210 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P398210 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98300 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81416 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897431 - -
          Affymetrix Rat Gene 1.0 ST Array 10897443 - -
          Affymetrix Rat Gene 1.0 ST Array 10897435 - -
          Affymetrix Rat Gene 1.0 ST Array 10897440 - -
          Affymetrix Rat Gene 1.0 ST Array 10897436 - -
          Affymetrix Rat Gene 1.0 ST Array 10897439 - -
          Affymetrix Rat Gene 1.0 ST Array 10897430 - -
          Affymetrix Rat Gene 1.0 ST Array 10897437 - -
          Affymetrix Rat Gene 1.0 ST Array 10897433 - -
          Affymetrix Rat Gene 1.0 ST Array 10897442 - -
          Affymetrix Rat Gene 1.0 ST Array 10897429 - -
          Affymetrix Rat Gene 1.0 ST Array 10897441 - -
          Affymetrix Rat Gene 1.0 ST Array 10897438 - -
          Affymetrix Rat Gene 1.0 ST Array 10897432 - -
          Affymetrix Rat Gene 1.1 ST Array 10897428 - -
          Rat Genome 230 2.0 Array 1369828_PM_at - -
          Rat Genome 230 2.0 Array 1369828_at - -
          Affymetrix HT Rat Focus Array 1369828_at - -
          Affymetrix HT Rat Focus Array 1369828_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700010443.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010444.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010447.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010440.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010450.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010451.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010449.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010452.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010438.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010441.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010445.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010448.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010439.rn.1 TC0700001411.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700010442.rn.1 TC0700001411.rn.1 RTA TC
          Rat Expression Set 230 1369828_at - -
          Rat Expression Set 230 1369828_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897428 - -
          Rat Genome U34 Set S79263_s_at - -
          Rat Genome U34 Set S79263_s_at - -
          Rat Genome U34 Set S79263_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21146582 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline