Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592259_m1
          See other PTPRH GE Assays ›
          Gene Symbol:
          Ptprh
          Gene Name:
          protein tyrosine phosphatase, receptor type, H
          Gene Aliases:
          Bem2
          Chromosome Location:
          Chr.1: 72809937 - 72859161 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592259_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptprh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191945.1 NP_001178874.1 11 - 12 1935 115
          XM_008758887.1 XP_008757109.1 14 - 15 3488 115
          GenBank mRNA D45413.1 - 3 - 4 484 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000865.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Protein Phosphatase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein phosphatase transmembrane signal receptor protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-tyrosine dephosphorylation
          transmembrane receptor protein tyrosine phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P290457 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290457 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P290457 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P290457 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P290457 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P489683 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701246124 - -
          Agilent Rat cDNA Microarray (G4105A) 701246124 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704030 - -
          Affymetrix Rat Gene 1.0 ST Array 10704028 - -
          Affymetrix Rat Gene 1.0 ST Array 10704021 - -
          Affymetrix Rat Gene 1.0 ST Array 10704025 - -
          Affymetrix Rat Gene 1.0 ST Array 10704026 - -
          Affymetrix Rat Gene 1.0 ST Array 10704029 - -
          Affymetrix Rat Gene 1.0 ST Array 10704032 - -
          Affymetrix Rat Gene 1.0 ST Array 10704020 - -
          Affymetrix Rat Gene 1.0 ST Array 10704027 - -
          Affymetrix Rat Gene 1.0 ST Array 10704022 - -
          Affymetrix Rat Gene 1.0 ST Array 10704033 - -
          Affymetrix Rat Gene 1.0 ST Array 10704031 - -
          Affymetrix Rat Gene 1.0 ST Array 10704023 - -
          Affymetrix Rat Gene 1.0 ST Array 10704024 - -
          Affymetrix Rat Gene 1.1 ST Array 10704018 - -
          Rat Genome 230 2.0 Array 1387444_PM_at - -
          Rat Genome 230 2.0 Array 1387444_at - -
          Affymetrix HT Rat Focus Array 1387444_PM_at - -
          Affymetrix HT Rat Focus Array 1387444_at - -
          Affymetrix Rat Clariom S Assay PSR0100005774.rn.1 TC0100000865.rn.1 RTA TC
          Rat Expression Set 230 1387444_at - -
          Rat Expression Set 230 1387444_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704018 - -
          Rat Genome U34 Set D45413_at - -
          Rat Genome U34 Set D45413_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21233018 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364327 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349302 - -
          RatRef-12 Expression BeadChip ILMN_48930 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline