Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592368_m1
          See other EHHADH GE Assays ›
          Gene Symbol:
          Ehhadh
          Gene Name:
          enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase
          Gene Aliases:
          1, Lbp, MEF, Mfe, Mfe1, Pbe, Pbfe, perMFE-1
          Chromosome Location:
          Chr.11: 82945104 - 82978364 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592368_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ehhadh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133606.1 NP_598290.1 6 - 7 928 53
          GenBank mRNA BC089777.1 - 6 - 7 917 7321434 53
          K03249.1 - 6 - 7 928 53

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000910.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ehhadh

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          internal protein amino acid acetylation
          fatty acid beta-oxidation
          3-hydroxyacyl-CoA dehydrogenase activity
          dodecenoyl-CoA delta-isomerase activity
          enoyl-CoA hydratase activity
          receptor binding
          enzyme binding
          coenzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58997 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124804 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13230 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13230 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13230 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13230 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701469162 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700935113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52680 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94231 - -
          Agilent Rat cDNA Microarray (G4105A) 701469162 - -
          Agilent Rat cDNA Microarray (G4105A) 700935113 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10752085 - -
          Affymetrix Rat Gene 1.0 ST Array 10752084 - -
          Affymetrix Rat Gene 1.0 ST Array 10752088 - -
          Affymetrix Rat Gene 1.0 ST Array 10752086 - -
          Affymetrix Rat Gene 1.0 ST Array 10752083 - -
          Affymetrix Rat Gene 1.0 ST Array 10752087 - -
          Affymetrix Rat Gene 1.0 ST Array 10752082 - -
          Affymetrix Rat Gene 1.1 ST Array 10752081 - -
          Rat Genome 230 2.0 Array 1368283_PM_at - -
          Rat Genome 230 2.0 Array 1368283_at - -
          Affymetrix HT Rat Focus Array 1368283_PM_at - -
          Affymetrix HT Rat Focus Array 1368283_at - -
          Affymetrix Rat Clariom S Assay PSR1100005154.rn.1 TC1100000910.rn.1 RTA TC
          Rat Expression Set 230 1368283_PM_at - -
          Rat Expression Set 230 1368283_at - -
          Affymetrix Rat Gene 1.0 ST Array 10752081 - -
          Rat Genome U34 Set K03249_at - -
          Rat Genome U34 Set M97197_at - -
          Rat Genome U34 Set K03249_at - -
          Rat Genome U34 Set M97197_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21936000 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline