Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592382_m1
          See other SLCO4A1 GE Assays ›
          Gene Symbol:
          Slco4a1
          Gene Name:
          solute carrier organic anion transporter family, member 4a1
          Gene Aliases:
          OATP-E, Slc21a12
          Chromosome Location:
          Chr.3: 175838994 - 175857985 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592382_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slco4a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133608.2 NP_598292.2 5 - 6 1289 81
          GenBank mRNA AF239262.1 - 4 - 5 1123 81
          BC097285.1 - 5 - 6 1289 7441748 81
          BC097286.1 - 5 - 6 1231 7444297 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002565.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slco4a1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          thyroid hormone metabolic process
          thyroid hormone transport
          transporter activity
          thyroid hormone transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P156840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78834 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156840 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P156840 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P156840 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842822 - -
          Affymetrix Rat Gene 1.0 ST Array 10842812 - -
          Affymetrix Rat Gene 1.0 ST Array 10842817 - -
          Affymetrix Rat Gene 1.0 ST Array 10842814 - -
          Affymetrix Rat Gene 1.0 ST Array 10842813 - -
          Affymetrix Rat Gene 1.0 ST Array 10842818 - -
          Affymetrix Rat Gene 1.0 ST Array 10842816 - -
          Affymetrix Rat Gene 1.0 ST Array 10842825 - -
          Affymetrix Rat Gene 1.0 ST Array 10842820 - -
          Affymetrix Rat Gene 1.0 ST Array 10842819 - -
          Affymetrix Rat Gene 1.0 ST Array 10842815 - -
          Affymetrix Rat Gene 1.0 ST Array 10842824 - -
          Affymetrix Rat Gene 1.0 ST Array 10842827 - -
          Affymetrix Rat Gene 1.0 ST Array 10842826 - -
          Affymetrix Rat Gene 1.1 ST Array 10842811 - -
          Rat Genome 230 2.0 Array 1389067_at - -
          Rat Genome 230 2.0 Array 1369437_PM_at - -
          Rat Genome 230 2.0 Array 1389067_PM_at - -
          Rat Genome 230 2.0 Array 1369437_at - -
          Affymetrix HT Rat Focus Array 1369437_at - -
          Affymetrix HT Rat Focus Array 1389067_PM_at - -
          Affymetrix HT Rat Focus Array 1389067_at - -
          Affymetrix HT Rat Focus Array 1369437_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300018875.rn.1 TC0300002565.rn.1 RTA TC
          Rat Expression Set 230 1369437_at - -
          Rat Expression Set 230 1369437_PM_at - -
          Rat Expression Set 230 1389067_at - -
          Rat Expression Set 230 1389067_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842811 - -
          Rat Genome U34 Set rc_AI180349_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21923642 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373636 - -
          RatRef-12 Expression BeadChip ILMN_65142 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline