Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592446_m1
          See other HOPX GE Assays ›
          Gene Symbol:
          Hopx
          Gene Name:
          HOP homeobox
          Gene Aliases:
          GIIg15b, Hod, Obl
          Chromosome Location:
          Chr.14: 33354803 - 33362547 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592446_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hopx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133621.2 NP_598305.2 2 - 3 259 77
          GenBank mRNA AF474162.1 - 179 77
          AF492685.1 - 2 - 3 259 77
          BC070950.1 - 1 - 2 170 7098349 77
          FQ212752.1 - 2 - 3 256 77
          FQ220854.1 - 2 - 3 258 77
          FQ221225.1 - 2 - 3 274 77
          FQ222004.1 - 2 - 3 249 77
          FQ222724.1 - 2 - 3 191 77
          FQ229425.1 - 2 - 3 244 77
          FQ229457.1 - 2 - 3 236 77
          FQ229504.1 - 2 - 3 236 77
          FQ229663.1 - 2 - 3 248 77
          FQ229876.1 - 2 - 3 236 77
          FQ229892.1 - 2 - 3 274 77
          FQ230105.1 - 1 - 2 252 77
          FQ230629.1 - 2 - 3 257 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000378.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hopx

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          trophectodermal cell differentiation
          transcription, DNA-templated
          heart development
          regulation of heart contraction
          histone deacetylation
          regulation of protein binding
          positive regulation of skeletal muscle tissue regeneration
          negative regulation of cell differentiation
          lung alveolus development
          positive regulation of striated muscle cell differentiation
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P404591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116508 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P404591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P404591 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P404591 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P404591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2394 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70508 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772199 - -
          Affymetrix Rat Gene 1.0 ST Array 10772195 - -
          Affymetrix Rat Gene 1.0 ST Array 10772198 - -
          Affymetrix Rat Gene 1.0 ST Array 10772196 - -
          Affymetrix Rat Gene 1.1 ST Array 10772194 - -
          Rat Genome 230 2.0 Array 1367816_at - -
          Rat Genome 230 2.0 Array 1367816_PM_at - -
          Affymetrix HT Rat Focus Array 1367816_PM_at - -
          Affymetrix HT Rat Focus Array 1367816_at - -
          Affymetrix Rat Clariom S Assay PSR1400002963.rn.1 TC1400000378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002969.rn.1 TC1400000378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002967.rn.1 TC1400000378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002968.rn.1 TC1400000378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002965.rn.1 TC1400000378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400002966.rn.1 TC1400000378.rn.1 RTA TC
          Rat Expression Set 230 1367816_at - -
          Rat Expression Set 230 1367816_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10772194 - -
          Rat Genome U34 Set rc_AI102739_at - -
          Rat Genome U34 Set rc_AI102887_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22336986 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363702 - -
          RatRef-12 Expression BeadChip ILMN_51495 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline