Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592480_m1
          See other ALB GE Assays ›
          Gene Symbol:
          Alb
          Gene Name:
          albumin
          Gene Aliases:
          Alb1, Albza
          Chromosome Location:
          Chr.14: 19176275 - 19191793 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592480_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134326.2 NP_599153.2 6 - 7 729 96
          GenBank mRNA BC085359.1 - 6 - 7 729 7303856 96
          FQ209382.1 - 2 - 3 177 96
          FQ209695.1 - 2 - 3 163 96
          FQ209784.1 - 6 - 7 748 96
          FQ209907.1 - 2 - 3 198 96
          FQ209957.1 - 1 - 2 110 96
          FQ210057.1 - 2 - 3 128 96
          FQ210436.1 - 6 - 7 748 96
          FQ210724.1 - 3 - 4 253 96
          FQ210803.1 - 2 - 3 199 96
          FQ210850.1 - 1 - 2 45 96
          FQ210851.1 - 1 - 2 76 96
          FQ210858.1 - 3 - 4 283 96
          FQ211218.1 - 3 - 4 357 96
          FQ211255.1 - 2 - 3 177 96
          FQ211259.1 - 2 - 3 186 96
          FQ218125.1 - 6 - 7 748 96
          FQ218216.1 - 1 - 2 53 96
          FQ218458.1 - 2 - 3 193 96
          FQ218739.1 - 1 - 2 98 96
          FQ218815.1 - 1 - 2 63 96
          FQ219210.1 - 2 - 3 148 96
          FQ219293.1 - 2 - 3 177 96
          FQ219447.1 - 2 - 3 170 96
          FQ219470.1 - 1 - 2 48 96
          FQ219670.1 - 2 - 3 227 96
          V01222.1 - 6 - 7 714 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001516.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Hypoxia Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Alb

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          transport
          response to nutrient
          cellular response to starvation
          response to organic substance
          hemolysis by symbiont of host erythrocytes
          vasodilation
          negative regulation of apoptotic process
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          response to mercury ion
          maintenance of mitochondrion location
          response to platinum ion
          DNA binding
          fatty acid binding
          drug binding
          zinc ion binding
          toxic substance binding
          oxygen binding
          enzyme binding
          pyridoxal phosphate binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1017367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50743 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11873 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1017367 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1017367 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1017367 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468107 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1017367 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468103 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701884483 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701431070 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2465 - -
          Agilent Rat cDNA Microarray (G4105A) 701431070 - -
          Agilent Rat cDNA Microarray (G4105A) 701884483 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775981 - -
          Affymetrix Rat Gene 1.0 ST Array 10775973 - -
          Affymetrix Rat Gene 1.0 ST Array 10775983 - -
          Affymetrix Rat Gene 1.0 ST Array 10775974 - -
          Affymetrix Rat Gene 1.0 ST Array 10775980 - -
          Affymetrix Rat Gene 1.0 ST Array 10775972 - -
          Affymetrix Rat Gene 1.0 ST Array 10775976 - -
          Affymetrix Rat Gene 1.0 ST Array 10775978 - -
          Affymetrix Rat Gene 1.0 ST Array 10775971 - -
          Affymetrix Rat Gene 1.0 ST Array 10775969 - -
          Affymetrix Rat Gene 1.0 ST Array 10775979 - -
          Affymetrix Rat Gene 1.0 ST Array 10775982 - -
          Affymetrix Rat Gene 1.0 ST Array 10775977 - -
          Affymetrix Rat Gene 1.0 ST Array 10775970 - -
          Affymetrix Rat Gene 1.0 ST Array 10775975 - -
          Affymetrix Rat Gene 1.1 ST Array 10775968 - -
          Rat Genome 230 2.0 Array 1367555_PM_at - -
          Rat Genome 230 2.0 Array 1367556_PM_s_at - -
          Rat Genome 230 2.0 Array 1367555_at - -
          Rat Genome 230 2.0 Array 1367556_s_at - -
          Affymetrix HT Rat Focus Array 1367556_s_at - -
          Affymetrix HT Rat Focus Array 1367555_PM_at - -
          Affymetrix HT Rat Focus Array 1367555_at - -
          Affymetrix HT Rat Focus Array 1367556_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR1400011181.rn.1 TC1400001516.rn.1 RTA TC
          Rat Expression Set 230 1367556_s_at - -
          Rat Expression Set 230 1367555_PM_at - -
          Rat Expression Set 230 1367555_at - -
          Rat Expression Set 230 1367556_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775968 - -
          Rat Genome U34 Set rc_AA860062_at - -
          Rat Genome U34 Set rc_AA945321_at - -
          Rat Genome U34 Set rc_AA866237_s_at - -
          Rat Genome U34 Set rc_AA860062_g_at - -
          Rat Genome U34 Set rc_AA818918_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21581708 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038793 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038792 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline