Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592499_m1
          See other ADH7 GE Assays ›
          Gene Symbol:
          Adh7
          Gene Name:
          alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 243500558 - 243516865 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592499_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adh7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134329.1 NP_599156.1 8 - 9 1159 98
          XM_008761493.1 XP_008759715.1 9 - 10 1546 98
          GenBank mRNA X98746.1 - 8 - 9 1159 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002288.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adh7

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          ethanol metabolic process
          ethanol catabolic process
          ethanol oxidation
          response to bacterium
          fatty acid omega-oxidation
          retinol metabolic process
          retinoic acid metabolic process
          response to ethanol
          oxidation-reduction process
          extracellular negative regulation of signal transduction
          alcohol dehydrogenase (NAD) activity
          alcohol dehydrogenase activity, zinc-dependent
          aldehyde oxidase activity
          retinol dehydrogenase activity
          zinc ion binding
          retinol binding
          ethanol binding
          protein homodimerization activity
          receptor antagonist activity
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P166161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P166161 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P166161 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P166161 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P166161 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700822807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57398 - -
          Agilent Rat cDNA Microarray (G4105A) 700822807 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819370 - -
          Affymetrix Rat Gene 1.0 ST Array 10819372 - -
          Affymetrix Rat Gene 1.0 ST Array 10819375 - -
          Affymetrix Rat Gene 1.0 ST Array 10819377 - -
          Affymetrix Rat Gene 1.0 ST Array 10819371 - -
          Affymetrix Rat Gene 1.0 ST Array 10819374 - -
          Affymetrix Rat Gene 1.0 ST Array 10819376 - -
          Affymetrix Rat Gene 1.0 ST Array 10819378 - -
          Affymetrix Rat Gene 1.0 ST Array 10819373 - -
          Affymetrix Rat Gene 1.1 ST Array 10819369 - -
          Rat Genome 230 2.0 Array 1369072_at - -
          Rat Genome 230 2.0 Array 1369072_PM_at - -
          Affymetrix HT Rat Focus Array 1369072_at - -
          Affymetrix HT Rat Focus Array 1369072_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200015943.rn.1 TC0200002288.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200015944.rn.1 TC0200002288.rn.1 RTA TC
          Rat Expression Set 230 1369072_at - -
          Rat Expression Set 230 1369072_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819369 - -
          Rat Genome U34 Set X98746cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21308400 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline