Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592680_m1
          See other ACMSD GE Assays ›
          Gene Symbol:
          Acmsd
          Gene Name:
          aminocarboxymuconate semialdehyde decarboxylase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 44424689 - 44468734 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592680_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acmsd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134372.1 NP_599199.1 4 - 5 487 72
          GenBank mRNA AB069781.1 - 4 - 5 487 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000193.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acmsd

          Panther Classification:

          Molecular Function -

          decarboxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          tryptophan metabolic process
          nicotinamide metabolic process
          quinolinate metabolic process
          aminocarboxymuconate-semialdehyde decarboxylase activity
          carboxy-lyase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155067 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13249 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13249 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13249 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72423 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41533 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71596 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81547 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767291 - -
          Affymetrix Rat Gene 1.0 ST Array 10767295 - -
          Affymetrix Rat Gene 1.0 ST Array 10767294 - -
          Affymetrix Rat Gene 1.0 ST Array 10767296 - -
          Affymetrix Rat Gene 1.0 ST Array 10767293 - -
          Affymetrix Rat Gene 1.0 ST Array 10767297 - -
          Affymetrix Rat Gene 1.0 ST Array 10767292 - -
          Affymetrix Rat Gene 1.0 ST Array 10767298 - -
          Affymetrix Rat Gene 1.1 ST Array 10767290 - -
          Rat Genome 230 2.0 Array 1370147_PM_at - -
          Rat Genome 230 2.0 Array 1370147_at - -
          Rat Genome 230 2.0 Array 1383111_PM_at - -
          Rat Genome 230 2.0 Array 1383111_at - -
          Affymetrix HT Rat Focus Array 1370147_PM_at - -
          Affymetrix HT Rat Focus Array 1370147_at - -
          Affymetrix Rat Clariom S Assay PSR1300000982.rn.1 TC1300000193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000980.rn.1 TC1300000193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000981.rn.1 TC1300000193.rn.1 RTA TC
          Rat Expression Set 230 1370147_PM_at - -
          Rat Expression Set 230 1370147_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767290 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21589904 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376844 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline