Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592930_m1
          See other BZW2 GE Assays ›
          Gene Symbol:
          Bzw2
          Gene Name:
          basic leucine zipper and W2 domains 2
          Gene Aliases:
          Hfb2
          Chromosome Location:
          Chr.6: 55586754 - 55647650 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592930_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bzw2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134402.3 NP_599229.1 6 - 7 679 62
          GenBank mRNA AF031483.1 - 6 - 7 707 62
          BC063149.1 - 6 - 7 683 6890706 62
          FQ225034.1 - 6 - 7 655 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002332.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bzw2

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s)

          nervous system development
          cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163517 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118986 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105718 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127402 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127402 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127402 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P261883 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700531857 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46605 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1530 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89131 - -
          Agilent Rat cDNA Microarray (G4105A) 700531857 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889696 - -
          Affymetrix Rat Gene 1.0 ST Array 10889685 - -
          Affymetrix Rat Gene 1.0 ST Array 10889686 - -
          Affymetrix Rat Gene 1.0 ST Array 10889688 - -
          Affymetrix Rat Gene 1.0 ST Array 10889689 - -
          Affymetrix Rat Gene 1.0 ST Array 10889694 - -
          Affymetrix Rat Gene 1.0 ST Array 10889687 - -
          Affymetrix Rat Gene 1.0 ST Array 10889691 - -
          Affymetrix Rat Gene 1.0 ST Array 10889690 - -
          Affymetrix Rat Gene 1.0 ST Array 10889695 - -
          Affymetrix Rat Gene 1.0 ST Array 10889693 - -
          Affymetrix Rat Gene 1.0 ST Array 10889697 - -
          Affymetrix Rat Gene 1.0 ST Array 10889692 - -
          Affymetrix Rat Gene 1.1 ST Array 10889684 - -
          Rat Genome 230 2.0 Array 1370258_PM_at - -
          Rat Genome 230 2.0 Array 1370258_at - -
          Affymetrix HT Rat Focus Array 1370258_PM_at - -
          Affymetrix HT Rat Focus Array 1370258_at - -
          Affymetrix Rat Clariom S Assay PSR0600016841.rn.1 TC0600002332.rn.1 RTA TC
          Rat Expression Set 230 1370258_at - -
          Rat Expression Set 230 1370258_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889684 - -
          Rat Genome U34 Set rc_AA997744_at - -
          Rat Genome U34 Set rc_AI230113_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21920240 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356421 - -
          RatRef-12 Expression BeadChip ILMN_56351 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline