Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592945_m1
          See other ABTB2 GE Assays ›
          Gene Symbol:
          Abtb2
          Gene Name:
          ankyrin repeat and BTB (POZ) domain containing 2
          Gene Aliases:
          Cca3
          Chromosome Location:
          Chr.3: 93494706 - 93648750 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abtb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134403.2 NP_599230.2 5 - 6 1959 107
          GenBank mRNA AB000216.1 - 5 - 6 1969 104
          CV110237.1 - 2 - 3 306 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001289.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abtb2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to toxic substance
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P087906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163337 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087906 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087906 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P087906 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P551842 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701298282 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41755 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57933 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37484 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52951 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36832 - -
          Agilent Rat cDNA Microarray (G4105A) 701298282 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10838181 - -
          Affymetrix Rat Gene 1.0 ST Array 10838177 - -
          Affymetrix Rat Gene 1.0 ST Array 10838180 - -
          Affymetrix Rat Gene 1.0 ST Array 10838176 - -
          Affymetrix Rat Gene 1.0 ST Array 10838185 - -
          Affymetrix Rat Gene 1.0 ST Array 10838173 - -
          Affymetrix Rat Gene 1.0 ST Array 10838172 - -
          Affymetrix Rat Gene 1.0 ST Array 10838186 - -
          Affymetrix Rat Gene 1.0 ST Array 10838187 - -
          Affymetrix Rat Gene 1.0 ST Array 10838178 - -
          Affymetrix Rat Gene 1.0 ST Array 10838174 - -
          Affymetrix Rat Gene 1.0 ST Array 10838183 - -
          Affymetrix Rat Gene 1.0 ST Array 10838188 - -
          Affymetrix Rat Gene 1.0 ST Array 10838182 - -
          Affymetrix Rat Gene 1.0 ST Array 10838184 - -
          Affymetrix Rat Gene 1.0 ST Array 10838179 - -
          Affymetrix Rat Gene 1.0 ST Array 10838171 - -
          Affymetrix Rat Gene 1.0 ST Array 10838175 - -
          Affymetrix Rat Gene 1.1 ST Array 10838170 - -
          Rat Genome 230 2.0 Array 1368111_at - -
          Rat Genome 230 2.0 Array 1368111_PM_at - -
          Affymetrix HT Rat Focus Array 1368111_at - -
          Affymetrix HT Rat Focus Array 1368111_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300008754.rn.1 TC0300001289.rn.1 RTA TC
          Rat Expression Set 230 1368111_PM_at - -
          Rat Expression Set 230 1368111_at - -
          Affymetrix Rat Gene 1.0 ST Array 10838170 - -
          Rat Genome U34 Set AB000216_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21531200 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352900 - -
          RatRef-12 Expression BeadChip ILMN_57432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline