Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593027_m1
          See other MARC2 GE Assays ›
          Gene Symbol:
          Marc2
          Gene Name:
          mitochondrial amidoxime reducing component 2
          Gene Aliases:
          Mg87, Mosc2, mARC1
          Chromosome Location:
          Chr.13: 102724266 - 102755460 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593027_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Marc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134410.2 NP_599237.1 4 - 5 769 74
          GenBank mRNA AF095741.1 - 4 - 5 770 74
          BC061734.1 - 4 - 5 796 5598768 74
          FQ211188.1 - 1 - 2 108 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Marc2

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          nitrate metabolic process
          oxidation-reduction process
          catalytic activity
          nitrate reductase activity
          oxidoreductase activity
          molybdenum ion binding
          pyridoxal phosphate binding
          molybdopterin cofactor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P231227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30182 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231227 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231227 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P231227 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P231227 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255373 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700313893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84896 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43056 - -
          Agilent Rat cDNA Microarray (G4105A) 700313893 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770512 - -
          Affymetrix Rat Gene 1.0 ST Array 10770514 - -
          Affymetrix Rat Gene 1.0 ST Array 10770516 - -
          Affymetrix Rat Gene 1.0 ST Array 10770515 - -
          Affymetrix Rat Gene 1.0 ST Array 10770513 - -
          Affymetrix Rat Gene 1.1 ST Array 10770511 - -
          Rat Genome 230 2.0 Array 1368071_at - -
          Rat Genome 230 2.0 Array 1368071_PM_at - -
          Affymetrix HT Rat Focus Array 1368071_at - -
          Affymetrix HT Rat Focus Array 1368071_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300016619.rn.1 TC1300002284.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016622.rn.1 TC1300002284.rn.1 RTA TC
          Rat Expression Set 230 1368071_at - -
          Rat Expression Set 230 1368071_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770511 - -
          Rat Genome U34 Set AF095741_at - -
          Rat Genome U34 Set AF095741_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21677393 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372217 - -
          RatRef-12 Expression BeadChip ILMN_60584 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline