Search
Search

Prkcdbp| Gene Symbol: Prkcdbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134449.1 | NP_604444.1 | 1 - 2 | 422 | 111 | |
| GenBank mRNA | BC101398.1 | - | 1 - 2 | 408 | 7933258 | 111 |
| D85435.1 | - | 1 - 2 | 422 | 111 | ||
| FQ220155.1 | - | 1 - 2 | 430 | 111 | ||
Set Membership: |
|