Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593178_m1
          See other LBR GE Assays ›
          Gene Symbol:
          Lbr
          Gene Name:
          lamin B receptor
          Gene Aliases:
          Nbp60
          Chromosome Location:
          Chr.13: 100431390 - 100450209 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593178_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lbr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134453.1 NP_604448.1 2 - 3 381 76
          GenBank mRNA AB002466.1 - 2 - 3 381 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002099.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lbr

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear envelope organization
          mitotic nuclear division
          oxidation-reduction process
          DNA binding
          nuclear localization sequence binding
          oxidoreductase activity, acting on the CH-CH group of donors, NAD or NADP as acceptor
          poly(A) RNA binding
          chaperone binding
          chromo shadow domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126710 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13274 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13274 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13274 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13274 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701597721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10384 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70226 - -
          Agilent Rat cDNA Microarray (G4105A) 701597721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770407 - -
          Affymetrix Rat Gene 1.0 ST Array 10770406 - -
          Affymetrix Rat Gene 1.0 ST Array 10770399 - -
          Affymetrix Rat Gene 1.0 ST Array 10770402 - -
          Affymetrix Rat Gene 1.0 ST Array 10770405 - -
          Affymetrix Rat Gene 1.0 ST Array 10770398 - -
          Affymetrix Rat Gene 1.0 ST Array 10770408 - -
          Affymetrix Rat Gene 1.0 ST Array 10770403 - -
          Affymetrix Rat Gene 1.0 ST Array 10770404 - -
          Affymetrix Rat Gene 1.0 ST Array 10770401 - -
          Affymetrix Rat Gene 1.0 ST Array 10770397 - -
          Affymetrix Rat Gene 1.0 ST Array 10770400 - -
          Affymetrix Rat Gene 1.1 ST Array 10770396 - -
          Rat Genome 230 2.0 Array 1370136_at - -
          Rat Genome 230 2.0 Array 1370136_PM_at - -
          Affymetrix HT Rat Focus Array 1370136_PM_at - -
          Affymetrix HT Rat Focus Array 1370136_at - -
          Affymetrix Rat Clariom S Assay PSR1300014988.rn.1 TC1300002099.rn.1 RTA TC
          Rat Expression Set 230 1370136_PM_at - -
          Rat Expression Set 230 1370136_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770396 - -
          Rat Genome U34 Set rc_AI145490_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21718736 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0