Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593266_m1
          See other RNF38 GE Assays ›
          Gene Symbol:
          Rnf38
          Gene Name:
          ring finger protein 38
          Gene Aliases:
          Oip1
          Chromosome Location:
          Chr.5: 59606754 - 59713108 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593266_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134467.1 NP_604462.1 4 - 5 549 85
          XM_006238012.2 XP_006238074.1 4 - 5 799 85
          XM_006238014.2 XP_006238076.1 4 - 5 697 85
          XM_006238015.2 XP_006238077.1 3 - 4 649 85
          XM_006238016.2 XP_006238078.1 3 - 4 896 85
          XM_006238017.2 XP_006238079.1 3 - 4 1669 85
          XM_006238018.2 XP_006238080.1 4 - 5 737 85
          XM_006238020.2 XP_006238082.1 4 - 5 542 85
          XM_008763664.1 XP_008761886.1 3 - 4 1323 85
          XM_008763665.1 XP_008761887.1 3 - 4 1236 85
          XM_008763666.1 XP_008761888.1 3 - 4 1221 85
          XM_008763667.1 XP_008761889.1 5 - 6 692 85
          XM_008763668.1 XP_008761890.1 4 - 5 635 85
          GenBank mRNA AF480444.1 - 4 - 5 549 85
          BC083548.1 - 3 - 4 464 7134112 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002640.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf38

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          male gonad development
          protein ubiquitination
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P224326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33264 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107792 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P224326 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P224326 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P224326 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P224326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44443 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57924 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876482 - -
          Affymetrix Rat Gene 1.0 ST Array 10876485 - -
          Affymetrix Rat Gene 1.0 ST Array 10876488 - -
          Affymetrix Rat Gene 1.0 ST Array 10876486 - -
          Affymetrix Rat Gene 1.0 ST Array 10876480 - -
          Affymetrix Rat Gene 1.0 ST Array 10876489 - -
          Affymetrix Rat Gene 1.0 ST Array 10876484 - -
          Affymetrix Rat Gene 1.0 ST Array 10876490 - -
          Affymetrix Rat Gene 1.0 ST Array 10876481 - -
          Affymetrix Rat Gene 1.0 ST Array 10876491 - -
          Affymetrix Rat Gene 1.0 ST Array 10876487 - -
          Affymetrix Rat Gene 1.0 ST Array 10876483 - -
          Affymetrix Rat Gene 1.1 ST Array 10876479 - -
          Rat Genome 230 2.0 Array 1387329_at - -
          Rat Genome 230 2.0 Array 1387329_PM_at - -
          Affymetrix HT Rat Focus Array 1387329_PM_at - -
          Affymetrix HT Rat Focus Array 1387329_at - -
          Affymetrix Rat Clariom S Assay PSR0500019696.rn.1 TC0500002640.rn.1 RTA TC
          Rat Expression Set 230 1387329_PM_at - -
          Rat Expression Set 230 1387329_at - -
          Affymetrix Rat Gene 1.0 ST Array 10876479 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21575500 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365393 - -
          RatRef-12 Expression BeadChip ILMN_54966 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline