Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593303_m1
          See other OSGIN1 GE Assays ›
          Gene Symbol:
          Osgin1
          Gene Name:
          oxidative stress induced growth inhibitor 1
          Gene Aliases:
          Okl38
          Chromosome Location:
          Chr.19: 52056816 - 52085491 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593303_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Osgin1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138504.2 NP_612513.1 4 - 5 609 69
          XM_006255706.2 XP_006255768.1 7 - 8 668 69
          XM_006255707.2 XP_006255769.1 3 - 4 704 69
          XM_008772594.1 XP_008770816.1 2 - 3 530 69
          GenBank mRNA AF549441.1 - 4 - 5 609 69
          AF549442.1 - 3 - 4 510 69
          AY081218.1 - 2 - 3 258 69
          BC091111.1 - 4 - 5 605 7106386 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000708.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Osgin1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell growth
          positive regulation of apoptotic process
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P529581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86725 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P529581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P529581 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P529581 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P529581 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62579 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76698 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 549 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10808304 - -
          Affymetrix Rat Gene 1.0 ST Array 10808311 - -
          Affymetrix Rat Gene 1.0 ST Array 10808306 - -
          Affymetrix Rat Gene 1.0 ST Array 10808308 - -
          Affymetrix Rat Gene 1.0 ST Array 10808310 - -
          Affymetrix Rat Gene 1.0 ST Array 10808309 - -
          Affymetrix Rat Gene 1.0 ST Array 10808307 - -
          Affymetrix Rat Gene 1.0 ST Array 10808303 - -
          Affymetrix Rat Gene 1.1 ST Array 10808302 - -
          Rat Genome 230 2.0 Array 1370336_at - -
          Rat Genome 230 2.0 Array 1370336_PM_at - -
          Affymetrix HT Rat Focus Array 1370336_at - -
          Affymetrix HT Rat Focus Array 1370336_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900005855.rn.1 TC1900000708.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900005854.rn.1 TC1900000708.rn.1 RTA TC
          Rat Expression Set 230 1370336_PM_at - -
          Rat Expression Set 230 1370336_at - -
          Affymetrix Rat Gene 1.0 ST Array 10808302 - -
          Rat Genome U34 Set rc_AI236754_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21310129 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354748 - -
          RatRef-12 Expression BeadChip ILMN_60586 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline