Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593492_m1
          See other STAMBP GE Assays ›
          Gene Symbol:
          Stambp
          Gene Name:
          Stam binding protein
          Gene Aliases:
          Amsh
          Chromosome Location:
          Chr.4: 115249343 - 115277341 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593492_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stambp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138531.2 NP_612540.1 6 - 7 971 60
          XM_006236726.2 XP_006236788.1 7 - 8 1638 60
          XM_006236727.2 XP_006236789.1 8 - 9 1756 60
          XM_008763045.1 XP_008761267.1 6 - 7 3299 60
          XM_008763046.1 XP_008761268.1 7 - 8 2319 60
          XM_008763047.1 XP_008761269.1 7 - 8 2736 60
          XM_008763048.1 XP_008761270.1 6 - 7 2618 60
          GenBank mRNA AY083159.1 - 5 - 6 795 60
          BC061711.1 - 6 - 7 971 5597943 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003185.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stambp

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          proteolysis
          biological_process
          negative regulation of phosphatidylinositol 3-kinase signaling
          protein deubiquitination
          negative regulation of neuron apoptotic process
          negative regulation of Ras protein signal transduction
          molecular_function
          thiol-dependent ubiquitin-specific protease activity
          metallopeptidase activity
          protein domain specific binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104343 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137828 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P634825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78688 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863567 - -
          Affymetrix Rat Gene 1.0 ST Array 10863560 - -
          Affymetrix Rat Gene 1.0 ST Array 10863565 - -
          Affymetrix Rat Gene 1.0 ST Array 10863563 - -
          Affymetrix Rat Gene 1.0 ST Array 10863562 - -
          Affymetrix Rat Gene 1.0 ST Array 10863564 - -
          Affymetrix Rat Gene 1.0 ST Array 10863569 - -
          Affymetrix Rat Gene 1.0 ST Array 10863566 - -
          Affymetrix Rat Gene 1.0 ST Array 10863568 - -
          Affymetrix Rat Gene 1.0 ST Array 10863561 - -
          Affymetrix Rat Gene 1.1 ST Array 10863559 - -
          Rat Genome 230 2.0 Array 1370476_at - -
          Rat Genome 230 2.0 Array 1370476_PM_at - -
          Affymetrix HT Rat Focus Array 1370476_PM_at - -
          Affymetrix HT Rat Focus Array 1370476_at - -
          Affymetrix Rat Clariom S Assay PSR0400022627.rn.1 TC0400003185.rn.1 RTA TC
          Rat Expression Set 230 1370476_PM_at - -
          Rat Expression Set 230 1370476_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863559 - -
          Rat Genome U34 Set rc_H33725_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21897732 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366608 - -
          RatRef-12 Expression BeadChip ILMN_55048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline