Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593702_m1
          See other SNRK GE Assays ›
          Gene Symbol:
          Snrk
          Gene Name:
          SNF related kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 130749857 - 130805225 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593702_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snrk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138833.1 NP_620188.1 2 - 3 736 73
          XM_006244057.2 XP_006244119.1 3 - 4 782 73
          XM_006244058.2 XP_006244120.1 3 - 4 816 73
          XM_006244059.1 XP_006244121.1 3 - 4 775 73
          XM_006244060.2 XP_006244122.1 2 - 3 790 73
          XM_006244061.1 XP_006244123.1 2 - 3 796 73
          XM_006244062.2 XP_006244124.1 2 - 3 724 73
          XM_008766666.1 XP_008764888.1 2 - 3 715 73
          XM_008766667.1 XP_008764889.1 2 - 3 857 73
          GenBank mRNA X89383.1 - 2 - 3 736 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001799.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snrk

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of neuron apoptotic process
          magnesium ion binding
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049633 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37146 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P466230 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P466230 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P466230 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049633 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P466230 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77410 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86032 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10914064 - -
          Affymetrix Rat Gene 1.0 ST Array 10939173 - -
          Affymetrix Rat Gene 1.0 ST Array 10914063 - -
          Affymetrix Rat Gene 1.0 ST Array 10914065 - -
          Affymetrix Rat Gene 1.0 ST Array 10939172 - -
          Affymetrix Rat Gene 1.0 ST Array 10914062 - -
          Affymetrix Rat Gene 1.1 ST Array 10914061 - -
          Affymetrix Rat Gene 1.1 ST Array 10939168 - -
          Rat Genome 230 2.0 Array 1379662_PM_a_at - -
          Rat Genome 230 2.0 Array 1386282_PM_x_at - -
          Rat Genome 230 2.0 Array 1371066_PM_at - -
          Rat Genome 230 2.0 Array 1394163_PM_at - -
          Rat Genome 230 2.0 Array 1379662_a_at - -
          Rat Genome 230 2.0 Array 1394163_at - -
          Rat Genome 230 2.0 Array 1386282_x_at - -
          Rat Genome 230 2.0 Array 1371066_at - -
          Affymetrix HT Rat Focus Array 1371066_at - -
          Affymetrix HT Rat Focus Array 1371066_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800014394.rn.1 TC0800001799.rn.1 RTA TC
          Rat Expression Set 230 1371066_PM_at - -
          Rat Expression Set 230 1371066_at - -
          Rat Expression Set 230 1386282_x_at - -
          Rat Expression Set 230 1386282_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10939168 - -
          Rat Genome U34 Set X89383_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21589997 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376628 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline