Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593730_m1
          See other TGOLN2 GE Assays ›
          Gene Symbol:
          Tgoln2
          Gene Name:
          trans-golgi network protein 2
          Gene Aliases:
          Tgn38, Tgoln1, Ttgn1
          Chromosome Location:
          Chr.4: 100475415 - 100483168 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593730_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgoln2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138840.2 NP_620195.1 2 - 3 990 73
          GenBank mRNA BC098512.1 - 2 - 3 990 7382894 73
          X53565.1 - 2 - 3 988 73
          X64600.1 - 2 - 3 988 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003086.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgoln2

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          Golgi to endosome transport
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5052 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120437 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120437 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P217452 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13304 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700599742 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700252350 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700625148 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701394567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46861 - -
          Agilent Rat cDNA Microarray (G4105A) 700920741 - -
          Agilent Rat cDNA Microarray (G4105A) 701394567 - -
          Agilent Rat cDNA Microarray (G4105A) 700599742 - -
          Agilent Rat cDNA Microarray (G4105A) 700252350 - -
          Agilent Rat cDNA Microarray (G4105A) 700625148 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863272 - -
          Affymetrix Rat Gene 1.0 ST Array 10863271 - -
          Affymetrix Rat Gene 1.0 ST Array 10863274 - -
          Affymetrix Rat Gene 1.0 ST Array 10863270 - -
          Affymetrix Rat Gene 1.0 ST Array 10863273 - -
          Affymetrix Rat Gene 1.1 ST Array 10863269 - -
          Rat Genome 230 2.0 Array 1392159_PM_at - -
          Rat Genome 230 2.0 Array 1389965_at - -
          Rat Genome 230 2.0 Array 1371028_PM_at - -
          Rat Genome 230 2.0 Array 1387755_s_at - -
          Rat Genome 230 2.0 Array 1371028_at - -
          Rat Genome 230 2.0 Array 1387755_PM_s_at - -
          Rat Genome 230 2.0 Array 1389965_PM_at - -
          Rat Genome 230 2.0 Array 1392159_at - -
          Affymetrix HT Rat Focus Array 1387755_PM_s_at - -
          Affymetrix HT Rat Focus Array 1389965_at - -
          Affymetrix HT Rat Focus Array 1387755_s_at - -
          Affymetrix HT Rat Focus Array 1371028_PM_at - -
          Affymetrix HT Rat Focus Array 1389965_PM_at - -
          Affymetrix HT Rat Focus Array 1371028_at - -
          Affymetrix Rat Clariom S Assay PSR0400021932.rn.1 TC0400003086.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021931.rn.1 TC0400003086.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021929.rn.1 TC0400003086.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400021933.rn.1 TC0400003086.rn.1 RTA TC
          Rat Expression Set 230 1389965_at - -
          Rat Expression Set 230 1387755_PM_s_at - -
          Rat Expression Set 230 1387755_s_at - -
          Rat Expression Set 230 1371028_at - -
          Rat Expression Set 230 1371028_PM_at - -
          Rat Expression Set 230 1389965_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863269 - -
          Rat Genome U34 Set X53565_at - -
          Rat Genome U34 Set rc_AA926292_s_at - -
          Rat Genome U34 Set rc_AI104342_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21975459 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357907 - -
          RatRef-12 Expression BeadChip ILMN_59168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline