Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593794_m1
          See other GPM6B GE Assays ›
          Gene Symbol:
          Gpm6b
          Gene Name:
          glycoprotein m6b
          Gene Aliases:
          M6b
          Chromosome Location:
          Chr.X: 29604607 - 29751140 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593794_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpm6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138846.1 NP_620201.1 4 - 5 817 71
          XM_006256849.2 XP_006256911.1 5 - 6 1356 71
          XM_006256850.2 XP_006256912.1 5 - 6 1358 71
          XM_006256851.2 XP_006256913.1 4 - 5 880 71
          XM_008773151.1 XP_008771373.1 4 - 5 869 71
          GenBank mRNA AB036421.1 - 4 - 5 817 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001617.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpm6b

          Panther Classification:

          Molecular Function -

          myelin protein

          Gene Ontology Categories:

          Function(s)

          ossification
          nervous system development
          protein transport
          cell differentiation
          positive regulation of bone mineralization
          regulation of actin cytoskeleton organization
          negative regulation of serotonin uptake
          regulation of focal adhesion assembly
          extracellular matrix assembly
          negative regulation of protein localization to cell surface

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13307 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13307 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13307 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13307 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700478715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1939 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100866 - -
          Agilent Rat cDNA Microarray (G4105A) 700478715 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937676 - -
          Affymetrix Rat Gene 1.0 ST Array 10937669 - -
          Affymetrix Rat Gene 1.0 ST Array 10937668 - -
          Affymetrix Rat Gene 1.0 ST Array 10937677 - -
          Affymetrix Rat Gene 1.0 ST Array 10937670 - -
          Affymetrix Rat Gene 1.0 ST Array 10937672 - -
          Affymetrix Rat Gene 1.0 ST Array 10937673 - -
          Affymetrix Rat Gene 1.0 ST Array 10937671 - -
          Affymetrix Rat Gene 1.1 ST Array 10937666 - -
          Rat Genome 230 2.0 Array 1381996_PM_at - -
          Rat Genome 230 2.0 Array 1370389_PM_at - -
          Rat Genome 230 2.0 Array 1370389_at - -
          Rat Genome 230 2.0 Array 1381996_at - -
          Affymetrix HT Rat Focus Array 1370389_at - -
          Affymetrix HT Rat Focus Array 1370389_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00010448.rn.1 TC0X00001617.rn.1 RTA TC
          Rat Expression Set 230 1370389_at - -
          Rat Expression Set 230 1370389_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937666 - -
          Rat Genome U34 Set rc_AA946489_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22156064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline