Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593883_m1
          See other PROM2 GE Assays ›
          Gene Symbol:
          Prom2
          Gene Name:
          prominin 2
          Gene Aliases:
          Prom-rp, Promrp, Trprp
          Chromosome Location:
          Chr.3: 120092564 - 120107496 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593883_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prom2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138857.1 NP_620212.1 16 - 17 2059 58
          XM_006234947.2 XP_006235009.1 16 - 17 2858 58
          GenBank mRNA AF486828.1 - 16 - 17 2059 58
          FQ229034.1 - 13 - 14 1604 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004108.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prom2

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          signal transduction
          positive regulation of cell projection organization
          negative regulation of pinocytosis
          regulation of clathrin-mediated endocytosis
          negative regulation of caveolin-mediated endocytosis
          transmembrane signaling receptor activity
          growth factor activity
          cholesterol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P530547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368168 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P530547 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P530547 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P530547 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P530547 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49106 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10849684 - -
          Affymetrix Rat Gene 1.0 ST Array 10849689 - -
          Affymetrix Rat Gene 1.0 ST Array 10849668 - -
          Affymetrix Rat Gene 1.0 ST Array 10849691 - -
          Affymetrix Rat Gene 1.0 ST Array 10849674 - -
          Affymetrix Rat Gene 1.0 ST Array 10849678 - -
          Affymetrix Rat Gene 1.0 ST Array 10849692 - -
          Affymetrix Rat Gene 1.0 ST Array 10849688 - -
          Affymetrix Rat Gene 1.0 ST Array 10849672 - -
          Affymetrix Rat Gene 1.0 ST Array 10849671 - -
          Affymetrix Rat Gene 1.0 ST Array 10849683 - -
          Affymetrix Rat Gene 1.0 ST Array 10849670 - -
          Affymetrix Rat Gene 1.0 ST Array 10849686 - -
          Affymetrix Rat Gene 1.0 ST Array 10849685 - -
          Affymetrix Rat Gene 1.0 ST Array 10849675 - -
          Affymetrix Rat Gene 1.0 ST Array 10849681 - -
          Affymetrix Rat Gene 1.0 ST Array 10849676 - -
          Affymetrix Rat Gene 1.0 ST Array 10849673 - -
          Affymetrix Rat Gene 1.0 ST Array 10849682 - -
          Affymetrix Rat Gene 1.0 ST Array 10849690 - -
          Affymetrix Rat Gene 1.0 ST Array 10849669 - -
          Affymetrix Rat Gene 1.0 ST Array 10849677 - -
          Affymetrix Rat Gene 1.0 ST Array 10849687 - -
          Affymetrix Rat Gene 1.0 ST Array 10849667 - -
          Affymetrix Rat Gene 1.0 ST Array 10849680 - -
          Affymetrix Rat Gene 1.1 ST Array 10849666 - -
          Rat Genome 230 2.0 Array 1369347_s_at - -
          Rat Genome 230 2.0 Array 1369347_PM_s_at - -
          Affymetrix HT Rat Focus Array 1369347_s_at - -
          Affymetrix HT Rat Focus Array 1369347_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0300031978.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031965.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031969.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031976.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031980.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031981.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031961.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031968.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031973.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031984.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031962.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031966.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031971.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031974.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031957.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031983.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031958.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031959.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031960.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031963.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031975.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031977.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031985.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031987.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031956.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031964.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031982.rn.1 TC0300004108.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300031972.rn.1 TC0300004108.rn.1 RTA TC
          Rat Expression Set 230 1369347_s_at - -
          Rat Expression Set 230 1369347_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10849666 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21709753 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372421 - -
          RatRef-12 Expression BeadChip ILMN_53975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline