Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593931_m1
          See other OPN4 GE Assays ›
          Gene Symbol:
          Opn4
          Gene Name:
          opsin 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 10943377 - 10952549 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593931_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Opn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138860.1 NP_620215.1 9 - 10 1588 69
          GenBank mRNA AY072689.1 - 9 - 10 1588 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001243.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Opn4

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          visual perception
          phototransduction
          detection of visible light
          rhodopsin mediated signaling pathway
          protein-chromophore linkage
          regulation of circadian rhythm
          rhythmic process
          11-cis retinal binding
          G-protein coupled photoreceptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P131151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132950 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131151 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131151 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P131151 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P825626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56521 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80624 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790511 - -
          Affymetrix Rat Gene 1.0 ST Array 10790518 - -
          Affymetrix Rat Gene 1.0 ST Array 10790522 - -
          Affymetrix Rat Gene 1.0 ST Array 10790517 - -
          Affymetrix Rat Gene 1.0 ST Array 10790521 - -
          Affymetrix Rat Gene 1.0 ST Array 10790513 - -
          Affymetrix Rat Gene 1.0 ST Array 10790515 - -
          Affymetrix Rat Gene 1.0 ST Array 10790519 - -
          Affymetrix Rat Gene 1.0 ST Array 10790516 - -
          Affymetrix Rat Gene 1.0 ST Array 10790514 - -
          Affymetrix Rat Gene 1.1 ST Array 10790510 - -
          Rat Genome 230 2.0 Array 1374302_PM_at - -
          Rat Genome 230 2.0 Array 1374302_at - -
          Affymetrix HT Rat Focus Array 1374302_at - -
          Affymetrix HT Rat Focus Array 1374302_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600009566.rn.1 TC1600001243.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009567.rn.1 TC1600001243.rn.1 RTA TC
          Rat Expression Set 230 1374302_PM_at - -
          Rat Expression Set 230 1374302_at - -
          Affymetrix Rat Gene 1.0 ST Array 10790510 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20953172 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370657 - -
          RatRef-12 Expression BeadChip ILMN_53275 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline