Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00593950_m1
          See other PTGR1 GE Assays ›
          Gene Symbol:
          Ptgr1
          Gene Name:
          prostaglandin reductase 1
          Gene Aliases:
          Dig1, Ltb4dh
          Chromosome Location:
          Chr.5: 76110753 - 76129361 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00593950_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptgr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138863.2 NP_620218.1 8 - 9 795 62
          GenBank mRNA BC089775.1 - 8 - 9 795 7307365 62
          FQ218841.1 - 8 - 9 814 62
          FQ220898.1 - 8 - 9 816 62
          FQ229147.1 - 8 - 9 811 62
          FQ229679.1 - 8 - 9 811 62
          U66322.2 - 7 - 8 760 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002841.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptgr1

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin metabolic process
          response to toxic substance
          oxidation-reduction process
          zinc ion binding
          2-alkenal reductase [NAD(P)] activity
          13-prostaglandin reductase activity
          15-oxoprostaglandin 13-oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P698240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P698240 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P698240 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P698240 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P698240 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702126464 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23350 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33756 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94700 - -
          Agilent Rat cDNA Microarray (G4105A) 702126464 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877133 - -
          Affymetrix Rat Gene 1.0 ST Array 10877134 - -
          Affymetrix Rat Gene 1.0 ST Array 10877135 - -
          Affymetrix Rat Gene 1.0 ST Array 10877132 - -
          Affymetrix Rat Gene 1.0 ST Array 10877131 - -
          Affymetrix Rat Gene 1.0 ST Array 10877137 - -
          Affymetrix Rat Gene 1.0 ST Array 10877136 - -
          Affymetrix Rat Gene 1.0 ST Array 10877139 - -
          Affymetrix Rat Gene 1.0 ST Array 10877138 - -
          Affymetrix Rat Gene 1.0 ST Array 10877140 - -
          Affymetrix Rat Gene 1.1 ST Array 10877130 - -
          Rat Genome 230 2.0 Array 1388102_PM_at - -
          Rat Genome 230 2.0 Array 1388102_at - -
          Affymetrix HT Rat Focus Array 1388102_PM_at - -
          Affymetrix HT Rat Focus Array 1388102_at - -
          Affymetrix Rat Clariom S Assay PSR0500021057.rn.1 TC0500002841.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021058.rn.1 TC0500002841.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021056.rn.1 TC0500002841.rn.1 RTA TC
          Rat Expression Set 230 1388102_PM_at - -
          Rat Expression Set 230 1388102_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877130 - -
          Rat Genome U34 Set U66322_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21212895 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349635 - -
          RatRef-12 Expression BeadChip ILMN_62869 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline