Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594102_m1
          See other AKR1D1 GE Assays ›
          Gene Symbol:
          Akr1d1
          Gene Name:
          aldo-keto reductase family 1, member D1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 65110706 - 65143930 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594102_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akr1d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138884.1 NP_620239.1 4 - 5 495 77
          XM_006236285.1 XP_006236347.1 4 - 5 515 77
          GenBank mRNA D17309.1 - 4 - 5 495 77
          FQ211413.1 - 4 - 5 513 77
          FQ219185.1 - 4 - 5 514 77
          FQ219421.1 - 4 - 5 458 77
          FQ219730.1 - 4 - 5 458 77
          S80431.1 - 4 - 5 467 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000650.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akr1d1

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          bile acid biosynthetic process
          cholesterol catabolic process
          digestion
          C21-steroid hormone metabolic process
          androgen metabolic process
          bile acid catabolic process
          oxidation-reduction process
          oxidoreductase activity
          3-oxo-5-beta-steroid 4-dehydrogenase activity
          delta4-3-oxosteroid 5beta-reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P354893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175746 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P354893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P354893 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P354893 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P430706 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P552817 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700932016 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701255251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39920 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32970 - -
          Agilent Rat cDNA Microarray (G4105A) 701255251 - -
          Agilent Rat cDNA Microarray (G4105A) 700932016 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854552 - -
          Affymetrix Rat Gene 1.0 ST Array 10854554 - -
          Affymetrix Rat Gene 1.0 ST Array 10854550 - -
          Affymetrix Rat Gene 1.0 ST Array 10854553 - -
          Affymetrix Rat Gene 1.0 ST Array 10854549 - -
          Affymetrix Rat Gene 1.0 ST Array 10854555 - -
          Affymetrix Rat Gene 1.0 ST Array 10854551 - -
          Affymetrix Rat Gene 1.0 ST Array 10854556 - -
          Affymetrix Rat Gene 1.0 ST Array 10854557 - -
          Affymetrix Rat Gene 1.1 ST Array 10854548 - -
          Rat Genome 230 2.0 Array 1398310_at - -
          Rat Genome 230 2.0 Array 1392384_s_at - -
          Rat Genome 230 2.0 Array 1398310_PM_at - -
          Rat Genome 230 2.0 Array 1392384_PM_s_at - -
          Affymetrix HT Rat Focus Array 1392384_s_at - -
          Affymetrix HT Rat Focus Array 1392384_PM_s_at - -
          Affymetrix HT Rat Focus Array 1398310_at - -
          Affymetrix HT Rat Focus Array 1398310_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400004658.rn.1 TC0400000650.rn.1 RTA TC
          Rat Expression Set 230 1398310_at - -
          Rat Expression Set 230 1398310_PM_at - -
          Rat Expression Set 230 1392384_s_at - -
          Rat Expression Set 230 1392384_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10854548 - -
          Rat Genome U34 Set D17309_at - -
          Rat Genome U34 Set S80431_s_at - -
          Rat Genome U34 Set D17309_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22065818 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376409 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline