Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594224_m1
          See other PLB1 GE Assays ›
          Gene Symbol:
          Plb1
          Gene Name:
          phospholipase B1
          Gene Aliases:
          Phlpb
          Chromosome Location:
          Chr.6: 25375699 - 25507108 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594224_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138898.1 NP_620253.1 20 - 21 1373 87
          GenBank mRNA D63648.1 - 20 - 21 1373 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001995.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plb1

          Panther Classification:

          Molecular Function -

          hydrolase lipase phospholipase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          phospholipid metabolic process
          lipid catabolic process
          retinol metabolic process
          positive regulation of acrosome reaction
          lysophospholipase activity
          phospholipase A2 activity
          lipase activity
          retinyl-palmitate esterase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175235 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13329 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13329 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13329 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13329 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13547 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68869 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3970 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10888756 - -
          Affymetrix Rat Gene 1.0 ST Array 10888724 - -
          Affymetrix Rat Gene 1.0 ST Array 10888726 - -
          Affymetrix Rat Gene 1.0 ST Array 10888707 - -
          Affymetrix Rat Gene 1.0 ST Array 10888714 - -
          Affymetrix Rat Gene 1.0 ST Array 10888740 - -
          Affymetrix Rat Gene 1.0 ST Array 10888735 - -
          Affymetrix Rat Gene 1.0 ST Array 10888733 - -
          Affymetrix Rat Gene 1.0 ST Array 10888719 - -
          Affymetrix Rat Gene 1.0 ST Array 10888721 - -
          Affymetrix Rat Gene 1.0 ST Array 10888725 - -
          Affymetrix Rat Gene 1.0 ST Array 10888706 - -
          Affymetrix Rat Gene 1.0 ST Array 10888743 - -
          Affymetrix Rat Gene 1.0 ST Array 10888709 - -
          Affymetrix Rat Gene 1.0 ST Array 10888715 - -
          Affymetrix Rat Gene 1.0 ST Array 10888738 - -
          Affymetrix Rat Gene 1.0 ST Array 10888734 - -
          Affymetrix Rat Gene 1.0 ST Array 10888736 - -
          Affymetrix Rat Gene 1.0 ST Array 10888720 - -
          Affymetrix Rat Gene 1.0 ST Array 10888703 - -
          Affymetrix Rat Gene 1.0 ST Array 10888751 - -
          Affymetrix Rat Gene 1.0 ST Array 10888731 - -
          Affymetrix Rat Gene 1.0 ST Array 10888745 - -
          Affymetrix Rat Gene 1.0 ST Array 10888754 - -
          Affymetrix Rat Gene 1.0 ST Array 10888729 - -
          Affymetrix Rat Gene 1.0 ST Array 10888722 - -
          Affymetrix Rat Gene 1.0 ST Array 10888716 - -
          Affymetrix Rat Gene 1.0 ST Array 10888723 - -
          Affymetrix Rat Gene 1.0 ST Array 10888712 - -
          Affymetrix Rat Gene 1.0 ST Array 10888732 - -
          Affymetrix Rat Gene 1.0 ST Array 10888711 - -
          Affymetrix Rat Gene 1.0 ST Array 10888747 - -
          Affymetrix Rat Gene 1.0 ST Array 10888717 - -
          Affymetrix Rat Gene 1.0 ST Array 10888755 - -
          Affymetrix Rat Gene 1.0 ST Array 10888727 - -
          Affymetrix Rat Gene 1.0 ST Array 10888749 - -
          Affymetrix Rat Gene 1.0 ST Array 10888737 - -
          Affymetrix Rat Gene 1.0 ST Array 10888750 - -
          Affymetrix Rat Gene 1.0 ST Array 10888713 - -
          Affymetrix Rat Gene 1.0 ST Array 10888752 - -
          Affymetrix Rat Gene 1.0 ST Array 10888702 - -
          Affymetrix Rat Gene 1.0 ST Array 10888753 - -
          Affymetrix Rat Gene 1.0 ST Array 10888746 - -
          Affymetrix Rat Gene 1.0 ST Array 10888742 - -
          Affymetrix Rat Gene 1.0 ST Array 10888757 - -
          Affymetrix Rat Gene 1.0 ST Array 10888718 - -
          Affymetrix Rat Gene 1.0 ST Array 10888704 - -
          Affymetrix Rat Gene 1.0 ST Array 10888748 - -
          Affymetrix Rat Gene 1.0 ST Array 10888710 - -
          Affymetrix Rat Gene 1.0 ST Array 10888741 - -
          Affymetrix Rat Gene 1.0 ST Array 10888708 - -
          Affymetrix Rat Gene 1.0 ST Array 10888730 - -
          Affymetrix Rat Gene 1.0 ST Array 10888728 - -
          Affymetrix Rat Gene 1.0 ST Array 10888744 - -
          Affymetrix Rat Gene 1.0 ST Array 10888705 - -
          Affymetrix Rat Gene 1.1 ST Array 10888701 - -
          Rat Genome 230 2.0 Array 1370447_PM_at - -
          Rat Genome 230 2.0 Array 1370447_at - -
          Affymetrix HT Rat Focus Array 1370447_PM_at - -
          Affymetrix HT Rat Focus Array 1370447_at - -
          Affymetrix Rat Clariom S Assay PSR0600014331.rn.1 TC0600001995.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600014334.rn.1 TC0600001995.rn.1 RTA TC
          Rat Expression Set 230 1370447_PM_at - -
          Rat Expression Set 230 1370447_at - -
          Affymetrix Rat Gene 1.0 ST Array 10888701 - -
          Rat Genome U34 Set rc_AI639174_s_at - -
          Rat Genome U34 Set E13935cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21833515 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365906 - -
          RatRef-12 Expression BeadChip ILMN_69604 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline