Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594479_m1
          See other WBP2 GE Assays ›
          Gene Symbol:
          Wbp2
          Gene Name:
          WW domain binding protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 104629568 - 104637903 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594479_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138975.1 NP_620431.1 2 - 3 174 65
          XM_006247705.2 XP_006247767.1 2 - 3 254 65
          GenBank mRNA AF499026.1 - 2 - 3 174 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004139.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wbp2

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of protein localization
          positive regulation of gene expression, epigenetic
          positive regulation of transcription from RNA polymerase II promoter
          cellular response to estrogen stimulus
          positive regulation of histone H3-K14 acetylation
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II transcription coactivator activity
          protein binding
          chromatin DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P342117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89608 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P342117 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P342117 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P342117 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342117 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44283 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10749116 - -
          Affymetrix Rat Gene 1.0 ST Array 10749113 - -
          Affymetrix Rat Gene 1.0 ST Array 10749109 - -
          Affymetrix Rat Gene 1.0 ST Array 10749110 - -
          Affymetrix Rat Gene 1.0 ST Array 10749114 - -
          Affymetrix Rat Gene 1.0 ST Array 10749115 - -
          Affymetrix Rat Gene 1.0 ST Array 10749111 - -
          Affymetrix Rat Gene 1.0 ST Array 10749112 - -
          Affymetrix Rat Gene 1.1 ST Array 10749108 - -
          Rat Genome 230 2.0 Array 1374394_at - -
          Rat Genome 230 2.0 Array 1369592_PM_at - -
          Rat Genome 230 2.0 Array 1369592_at - -
          Rat Genome 230 2.0 Array 1374394_PM_at - -
          Affymetrix HT Rat Focus Array 1369592_PM_at - -
          Affymetrix HT Rat Focus Array 1369592_at - -
          Affymetrix HT Rat Focus Array 1374394_PM_at - -
          Affymetrix HT Rat Focus Array 1374394_at - -
          Affymetrix Rat Clariom S Assay PSR1000037593.rn.1 TC1000004139.rn.1 RTA TC
          Rat Expression Set 230 1374394_PM_at - -
          Rat Expression Set 230 1374394_at - -
          Rat Expression Set 230 1369592_PM_at - -
          Rat Expression Set 230 1369592_at - -
          Affymetrix Rat Gene 1.0 ST Array 10749108 - -
          Rat Genome U34 Set rc_AA944155_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21410641 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352211 - -
          RatRef-12 Expression BeadChip ILMN_57984 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline