Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594626_m1
          See other CGREF1 GE Assays ›
          Gene Symbol:
          Cgref1
          Gene Name:
          cell growth regulator with EF hand domain 1
          Gene Aliases:
          Cgr11
          Chromosome Location:
          Chr.6: 26797126 - 26809132 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594626_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cgref1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139087.2 NP_620787.2 2 - 3 227 76
          GenBank mRNA FM040765.1 - 2 - 3 227 76
          FQ220335.1 - 2 - 3 157 76
          U66470.1 - 2 - 3 148 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000299.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cgref1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle arrest
          cell adhesion
          negative regulation of cell growth
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P398193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336386 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161688 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398193 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P398193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398193 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21841 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18343 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8897 - -
          Agilent Rat cDNA Microarray (G4105A) 700312679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10883217 - -
          Affymetrix Rat Gene 1.0 ST Array 10883215 - -
          Affymetrix Rat Gene 1.0 ST Array 10883216 - -
          Affymetrix Rat Gene 1.0 ST Array 10883213 - -
          Affymetrix Rat Gene 1.0 ST Array 10883218 - -
          Affymetrix Rat Gene 1.0 ST Array 10883214 - -
          Affymetrix Rat Gene 1.1 ST Array 10883212 - -
          Rat Genome 230 2.0 Array 1370361_PM_at - -
          Rat Genome 230 2.0 Array 1370361_at - -
          Affymetrix HT Rat Focus Array 1370361_PM_at - -
          Affymetrix HT Rat Focus Array 1370361_at - -
          Affymetrix Rat Clariom S Assay PSR0600001982.rn.1 TC0600000299.rn.1 RTA TC
          Rat Expression Set 230 1370361_PM_at - -
          Rat Expression Set 230 1370361_at - -
          Affymetrix Rat Gene 1.0 ST Array 10883212 - -
          Rat Genome U34 Set U66470_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349519 - -
          RatRef-12 Expression BeadChip ILMN_58442 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline