Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594657_m1
          See other ACVR1C GE Assays ›
          Gene Symbol:
          Acvr1c
          Gene Name:
          activin A receptor, type IC
          Gene Aliases:
          Alk-7, Alk7, habrec1
          Chromosome Location:
          Chr.3: 44272859 - 44342501 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594657_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acvr1c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139090.1 NP_620790.1 8 - 9 1501 62
          GenBank mRNA U35025.1 - 8 - 9 1386 62
          U69702.1 - 8 - 9 1501 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003154.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acvr1c

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          in utero embryonic development
          response to dietary excess
          protein phosphorylation
          apoptotic process
          transforming growth factor beta receptor complex assembly
          determination of left/right symmetry
          response to glucose
          response to organic cyclic compound
          lipid storage
          cerebellum development
          hippocampus development
          signal transduction by protein phosphorylation
          cell differentiation
          apoptotic nuclear changes
          midbrain development
          response to insulin
          activin receptor signaling pathway
          response to lipid
          nodal signaling pathway
          response to drug
          regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of insulin secretion
          negative regulation of trophoblast cell migration
          negative regulation of chorionic trophoblast cell proliferation
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          ATP binding
          activin receptor activity, type I
          growth factor binding
          nodal binding
          SMAD binding
          metal ion binding
          activin binding
          BMP receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042249 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P065868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155514 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58285 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121241 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61505 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042249 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P065868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042249 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042249 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P065868 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P339702 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95061 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17572 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2963 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845427 - -
          Affymetrix Rat Gene 1.0 ST Array 10845417 - -
          Affymetrix Rat Gene 1.0 ST Array 10845421 - -
          Affymetrix Rat Gene 1.0 ST Array 10845420 - -
          Affymetrix Rat Gene 1.0 ST Array 10845418 - -
          Affymetrix Rat Gene 1.0 ST Array 10845425 - -
          Affymetrix Rat Gene 1.0 ST Array 10845428 - -
          Affymetrix Rat Gene 1.0 ST Array 10845426 - -
          Affymetrix Rat Gene 1.0 ST Array 10845422 - -
          Affymetrix Rat Gene 1.0 ST Array 10845423 - -
          Affymetrix Rat Gene 1.0 ST Array 10845424 - -
          Affymetrix Rat Gene 1.0 ST Array 10845419 - -
          Affymetrix Rat Gene 1.1 ST Array 10845416 - -
          Rat Genome 230 2.0 Array 1370534_PM_at - -
          Rat Genome 230 2.0 Array 1370534_at - -
          Affymetrix HT Rat Focus Array 1370534_PM_at - -
          Affymetrix HT Rat Focus Array 1370534_at - -
          Affymetrix Rat Clariom S Assay PSR0300024346.rn.1 TC0300003154.rn.1 RTA TC
          Rat Expression Set 230 1370534_at - -
          Rat Expression Set 230 1370534_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845416 - -
          Rat Genome U34 Set U63318_s_at - -
          Rat Genome U34 Set U69702_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21422964 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367140 - -
          RatRef-12 Expression BeadChip ILMN_59595 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline