Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594720_m1
          See other DDX46 GE Assays ›
          Gene Symbol:
          Ddx46
          Gene Name:
          DEAD (Asp-Glu-Ala-Asp) box polypeptide 46
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 9475275 - 9519544 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594720_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddx46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139098.1 NP_620798.1 18 - 19 2589 94
          XM_006253601.2 XP_006253663.1 18 - 19 2619 94
          GenBank mRNA BC107590.1 - 18 - 19 2455 7461248 94
          U25746.1 - 18 - 19 2589 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001311.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddx46

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          helicase activity
          ATP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P335059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5149 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150295 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P335059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P335059 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P335059 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P335059 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701827395 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701745505 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700310332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80835 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100481 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51566 - -
          Agilent Rat cDNA Microarray (G4105A) 701745505 - -
          Agilent Rat cDNA Microarray (G4105A) 700310332 - -
          Agilent Rat cDNA Microarray (G4105A) 701827395 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797190 - -
          Affymetrix Rat Gene 1.0 ST Array 10797206 - -
          Affymetrix Rat Gene 1.0 ST Array 10797208 - -
          Affymetrix Rat Gene 1.0 ST Array 10797210 - -
          Affymetrix Rat Gene 1.0 ST Array 10797205 - -
          Affymetrix Rat Gene 1.0 ST Array 10797191 - -
          Affymetrix Rat Gene 1.0 ST Array 10797200 - -
          Affymetrix Rat Gene 1.0 ST Array 10797192 - -
          Affymetrix Rat Gene 1.0 ST Array 10797195 - -
          Affymetrix Rat Gene 1.0 ST Array 10797193 - -
          Affymetrix Rat Gene 1.0 ST Array 10797212 - -
          Affymetrix Rat Gene 1.0 ST Array 10797207 - -
          Affymetrix Rat Gene 1.0 ST Array 10797211 - -
          Affymetrix Rat Gene 1.0 ST Array 10797202 - -
          Affymetrix Rat Gene 1.0 ST Array 10797196 - -
          Affymetrix Rat Gene 1.0 ST Array 10797201 - -
          Affymetrix Rat Gene 1.0 ST Array 10797209 - -
          Affymetrix Rat Gene 1.0 ST Array 10797198 - -
          Affymetrix Rat Gene 1.0 ST Array 10797199 - -
          Affymetrix Rat Gene 1.0 ST Array 10797194 - -
          Affymetrix Rat Gene 1.0 ST Array 10797197 - -
          Affymetrix Rat Gene 1.0 ST Array 10797203 - -
          Affymetrix Rat Gene 1.0 ST Array 10797204 - -
          Affymetrix Rat Gene 1.1 ST Array 10797189 - -
          Rat Genome 230 2.0 Array 1387912_PM_at - -
          Rat Genome 230 2.0 Array 1387912_at - -
          Affymetrix HT Rat Focus Array 1387912_at - -
          Affymetrix HT Rat Focus Array 1387912_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700008991.rn.1 TC1700001311.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008989.rn.1 TC1700001311.rn.1 RTA TC
          Rat Expression Set 230 1387912_PM_at - -
          Rat Expression Set 230 1387912_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797189 - -
          Rat Genome U34 Set U25746_at - -
          Rat Genome U34 Set U25746_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22175144 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374807 - -
          RatRef-12 Expression BeadChip ILMN_50648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline