Search
Search

Slc25a3| Gene Symbol: Slc25a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270788.1 | NP_001257717.1 | 6 - 7 | 910 | 63 | |
| NM_139100.2 | NP_620800.2 | 6 - 7 | 907 | 63 | ||
| GenBank mRNA | BC070918.1 | - | 6 - 7 | 849 | 7104964 | 63 |
| FQ210190.1 | - | 6 - 7 | 861 | 63 | ||
| FQ213208.1 | - | 1 - 2 | 73 | 63 | ||
| FQ213570.1 | - | 1 - 2 | 73 | 63 | ||
| FQ213750.1 | - | 6 - 7 | 866 | 63 | ||
| FQ214401.1 | - | 1 - 2 | 73 | 63 | ||
| FQ214581.1 | - | 1 - 2 | 73 | 63 | ||
| FQ214698.1 | - | 6 - 7 | 864 | 63 | ||
| FQ215697.1 | - | 6 - 7 | 822 | 63 | ||
| FQ215957.1 | - | 6 - 7 | 866 | 63 | ||
| FQ216084.1 | - | 1 - 2 | 73 | 63 | ||
| FQ216349.1 | - | 6 - 7 | 863 | 63 | ||
| FQ216361.1 | - | 6 - 7 | 864 | 63 | ||
| FQ216409.1 | - | 6 - 7 | 871 | 63 | ||
| FQ216421.1 | - | 6 - 7 | 864 | 63 | ||
| FQ216474.1 | - | 6 - 7 | 872 | 63 | ||
| FQ216716.1 | - | 1 - 2 | 73 | 63 | ||
| FQ216978.1 | - | 1 - 2 | 73 | 63 | ||
| FQ217343.1 | - | 1 - 2 | 73 | 63 | ||
| FQ217883.1 | - | 1 - 2 | 91 | 63 | ||
| FQ220537.1 | - | 6 - 7 | 861 | 63 | ||
| FQ220626.1 | - | 6 - 7 | 848 | 63 | ||
| FQ222611.1 | - | 1 - 2 | 73 | 63 | ||
| FQ223916.1 | - | 6 - 7 | 864 | 63 | ||
| FQ224211.1 | - | 1 - 2 | 73 | 63 | ||
| FQ227446.1 | - | 6 - 7 | 861 | 63 | ||
| FQ229113.1 | - | 6 - 7 | 861 | 63 | ||
| FQ230627.1 | - | 6 - 7 | 863 | 63 | ||
| FQ232293.1 | - | 4 - 5 | 666 | 63 | ||
| FQ232835.1 | - | 6 - 7 | 857 | 63 | ||
| FQ234854.1 | - | 6 - 7 | 861 | 63 | ||
| M23984.1 | - | 5 - 6 | 826 | 63 | ||
Set Membership: |
|