Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595008_m1
          See other EHD4 GE Assays ›
          Gene Symbol:
          Ehd4
          Gene Name:
          EH-domain containing 4
          Gene Aliases:
          Past2
          Chromosome Location:
          Chr.3: 111877853 - 111941841 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595008_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ehd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139324.1 NP_647540.1 5 - 6 1113 75
          GenBank mRNA AY094093.1 - 5 - 6 1113 75
          BC083175.1 - 5 - 6 1095 7105816 75
          FQ232475.1 - 2 - 3 274 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003996.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ehd4

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          pinocytosis
          regulation of endocytosis
          endocytic recycling
          positive regulation of peptidyl-tyrosine phosphorylation
          protein homooligomerization
          cellular response to growth factor stimulus
          calcium ion binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P423613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128626 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33870 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P423613 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P423613 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P423613 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P423613 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59158 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29761 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91141 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848739 - -
          Affymetrix Rat Gene 1.0 ST Array 10848736 - -
          Affymetrix Rat Gene 1.0 ST Array 10848738 - -
          Affymetrix Rat Gene 1.0 ST Array 10848735 - -
          Affymetrix Rat Gene 1.0 ST Array 10848737 - -
          Affymetrix Rat Gene 1.0 ST Array 10848734 - -
          Affymetrix Rat Gene 1.1 ST Array 10848733 - -
          Rat Genome 230 2.0 Array 1374399_PM_at - -
          Rat Genome 230 2.0 Array 1375739_at - -
          Rat Genome 230 2.0 Array 1372106_at - -
          Rat Genome 230 2.0 Array 1372106_PM_at - -
          Rat Genome 230 2.0 Array 1375739_PM_at - -
          Rat Genome 230 2.0 Array 1374399_at - -
          Affymetrix HT Rat Focus Array 1375739_at - -
          Affymetrix HT Rat Focus Array 1374399_at - -
          Affymetrix HT Rat Focus Array 1372106_PM_at - -
          Affymetrix HT Rat Focus Array 1375739_PM_at - -
          Affymetrix HT Rat Focus Array 1374399_PM_at - -
          Affymetrix HT Rat Focus Array 1372106_at - -
          Affymetrix Rat Clariom S Assay PSR0300030374.rn.1 TC0300003996.rn.1 RTA TC
          Rat Expression Set 230 1372106_PM_at - -
          Rat Expression Set 230 1375739_PM_at - -
          Rat Expression Set 230 1374399_at - -
          Rat Expression Set 230 1374399_PM_at - -
          Rat Expression Set 230 1375739_at - -
          Rat Expression Set 230 1372106_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848733 - -
          Rat Genome U34 Set rc_AA799461_at - -
          Rat Genome U34 Set rc_AI233822_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21486738 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365515 - -
          RatRef-12 Expression BeadChip ILMN_51306 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0