Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595164_m1
          See other FMO4 GE Assays ›
          Gene Symbol:
          Fmo4
          Gene Name:
          flavin containing monooxygenase 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 80685385 - 80703575 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595164_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fmo4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144561.2 NP_653147.1 5 - 6 803 77
          NM_144562.1 NP_653148.1 3 - 4 408 77
          XM_006250144.2 XP_006250206.1 5 - 6 738 77
          GenBank mRNA AF458416.1 - 5 - 6 804 77
          AF458417.1 - 3 - 4 408 77
          BC081721.1 - 5 - 6 793 7107835 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001774.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fmo4

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          N,N-dimethylaniline monooxygenase activity
          flavin adenine dinucleotide binding
          NADP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P170368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119364 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170368 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P170368 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P170368 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P170368 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28657 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87285 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769352 - -
          Affymetrix Rat Gene 1.0 ST Array 10769360 - -
          Affymetrix Rat Gene 1.0 ST Array 10769351 - -
          Affymetrix Rat Gene 1.0 ST Array 10769350 - -
          Affymetrix Rat Gene 1.0 ST Array 10769353 - -
          Affymetrix Rat Gene 1.0 ST Array 10769357 - -
          Affymetrix Rat Gene 1.0 ST Array 10769358 - -
          Affymetrix Rat Gene 1.0 ST Array 10769356 - -
          Affymetrix Rat Gene 1.0 ST Array 10769355 - -
          Affymetrix Rat Gene 1.0 ST Array 10769354 - -
          Affymetrix Rat Gene 1.0 ST Array 10769359 - -
          Affymetrix Rat Gene 1.1 ST Array 10769349 - -
          Rat Genome 230 2.0 Array 1395026_at - -
          Rat Genome 230 2.0 Array 1379885_PM_at - -
          Rat Genome 230 2.0 Array 1395026_PM_at - -
          Rat Genome 230 2.0 Array 1379885_at - -
          Affymetrix HT Rat Focus Array 1395026_PM_at - -
          Affymetrix HT Rat Focus Array 1395026_at - -
          Affymetrix HT Rat Focus Array 1379885_PM_at - -
          Affymetrix HT Rat Focus Array 1379885_at - -
          Affymetrix Rat Clariom S Assay PSR1300012561.rn.1 TC1300001774.rn.1 RTA TC
          Rat Expression Set 230 1395026_PM_at - -
          Rat Expression Set 230 1395026_at - -
          Rat Expression Set 230 1379885_at - -
          Rat Expression Set 230 1379885_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769349 - -
          Rat Genome U34 Set rc_AI232461_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22320480 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376656 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375784 - -
          RatRef-12 Expression BeadChip ILMN_51217 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline