Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595193_m1
          See other PNPLA7 GE Assays ›
          Gene Symbol:
          Pnpla7
          Gene Name:
          patatin-like phospholipase domain containing 7
          Gene Aliases:
          Nte-related, Ntel, Ntel1
          Chromosome Location:
          Chr.3: 2182888 - 2261879 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595193_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pnpla7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144738.2 NP_653339.2 25 - 26 3088 67
          GenBank mRNA AY100477.1 - 7 - 8 890 67
          BC083547.1 - 8 - 9 894 7132749 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005070.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pnpla7

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          developmental process
          lysophospholipase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173251 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119567 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119567 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1000875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33817 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40199 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64769 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46115 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96373 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834503 - -
          Affymetrix Rat Gene 1.0 ST Array 10834536 - -
          Affymetrix Rat Gene 1.0 ST Array 10834513 - -
          Affymetrix Rat Gene 1.0 ST Array 10834516 - -
          Affymetrix Rat Gene 1.0 ST Array 10834518 - -
          Affymetrix Rat Gene 1.0 ST Array 10834509 - -
          Affymetrix Rat Gene 1.0 ST Array 10834523 - -
          Affymetrix Rat Gene 1.0 ST Array 10834514 - -
          Affymetrix Rat Gene 1.0 ST Array 10834531 - -
          Affymetrix Rat Gene 1.0 ST Array 10834519 - -
          Affymetrix Rat Gene 1.0 ST Array 10834515 - -
          Affymetrix Rat Gene 1.0 ST Array 10834504 - -
          Affymetrix Rat Gene 1.0 ST Array 10834508 - -
          Affymetrix Rat Gene 1.0 ST Array 10834517 - -
          Affymetrix Rat Gene 1.0 ST Array 10834535 - -
          Affymetrix Rat Gene 1.0 ST Array 10834527 - -
          Affymetrix Rat Gene 1.0 ST Array 10834521 - -
          Affymetrix Rat Gene 1.0 ST Array 10834533 - -
          Affymetrix Rat Gene 1.0 ST Array 10834534 - -
          Affymetrix Rat Gene 1.0 ST Array 10834529 - -
          Affymetrix Rat Gene 1.0 ST Array 10834522 - -
          Affymetrix Rat Gene 1.0 ST Array 10834540 - -
          Affymetrix Rat Gene 1.0 ST Array 10834520 - -
          Affymetrix Rat Gene 1.0 ST Array 10834532 - -
          Affymetrix Rat Gene 1.0 ST Array 10834525 - -
          Affymetrix Rat Gene 1.0 ST Array 10834538 - -
          Affymetrix Rat Gene 1.0 ST Array 10834505 - -
          Affymetrix Rat Gene 1.0 ST Array 10834539 - -
          Affymetrix Rat Gene 1.0 ST Array 10834528 - -
          Affymetrix Rat Gene 1.0 ST Array 10834510 - -
          Affymetrix Rat Gene 1.0 ST Array 10834507 - -
          Affymetrix Rat Gene 1.0 ST Array 10834530 - -
          Affymetrix Rat Gene 1.0 ST Array 10834506 - -
          Affymetrix Rat Gene 1.0 ST Array 10834526 - -
          Affymetrix Rat Gene 1.0 ST Array 10834541 - -
          Affymetrix Rat Gene 1.0 ST Array 10834524 - -
          Affymetrix Rat Gene 1.0 ST Array 10834511 - -
          Affymetrix Rat Gene 1.0 ST Array 10834537 - -
          Affymetrix Rat Gene 1.0 ST Array 10834512 - -
          Affymetrix Rat Gene 1.1 ST Array 10834502 - -
          Rat Genome 230 2.0 Array 1396684_PM_at - -
          Rat Genome 230 2.0 Array 1396684_at - -
          Rat Genome 230 2.0 Array 1377049_at - -
          Rat Genome 230 2.0 Array 1377049_PM_at - -
          Affymetrix HT Rat Focus Array 1377049_PM_at - -
          Affymetrix HT Rat Focus Array 1396684_PM_at - -
          Affymetrix HT Rat Focus Array 1396684_at - -
          Affymetrix HT Rat Focus Array 1377049_at - -
          Affymetrix Rat Clariom S Assay PSR0300038988.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038992.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038996.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039003.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039006.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039025.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039029.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038990.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039014.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039030.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038986.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038994.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039011.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038984.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038998.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039004.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039009.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039020.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039026.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039028.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039032.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038983.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038993.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038995.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038999.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039016.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038989.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039002.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039031.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038982.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038985.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038987.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038991.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038997.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039005.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039008.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039010.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039017.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039021.rn.1 TC0300005070.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300039023.rn.1 TC0300005070.rn.1 RTA TC
          Rat Expression Set 230 1377049_at - -
          Rat Expression Set 230 1377049_PM_at - -
          Rat Expression Set 230 1396684_at - -
          Rat Expression Set 230 1396684_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834502 - -
          Rat Genome U34 Set rc_AI639348_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20812969 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline