Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595250_m1
          See other ADIPOQ GE Assays ›
          Gene Symbol:
          Adipoq
          Gene Name:
          adiponectin, C1Q and collagen domain containing
          Gene Aliases:
          Acdc, Acrp30
          Chromosome Location:
          Chr.11: 81330845 - 81344488 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595250_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adipoq
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144744.3 NP_653345.1 2 - 3 301 63
          GenBank mRNA AY033885.1 - 2 - 3 248 63
          BC092565.1 - 1 - 2 237 7308618 63
          FM047633.1 - 2 - 3 301 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001880.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adipoq

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of protein phosphorylation
          glucose metabolic process
          fatty acid beta-oxidation
          response to nutrient
          circadian rhythm
          response to sucrose
          response to glucose
          positive regulation of signal transduction
          negative regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of protein kinase A signaling
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of cholesterol efflux
          regulation of glucose metabolic process
          response to activity
          negative regulation of smooth muscle cell migration
          fatty acid oxidation
          negative regulation of cell migration
          negative regulation of granulocyte differentiation
          response to nutrient levels
          negative regulation of protein autophosphorylation
          positive regulation of cellular protein metabolic process
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-8 production
          cellular response to insulin stimulus
          positive regulation of myeloid cell apoptotic process
          adiponectin-activated signaling pathway
          negative regulation of heterotypic cell-cell adhesion
          low-density lipoprotein particle clearance
          response to tumor necrosis factor
          cellular response to drug
          response to drug
          glucose homeostasis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of MAP kinase activity
          response to ethanol
          negative regulation of fat cell differentiation
          negative regulation of macrophage differentiation
          negative regulation of low-density lipoprotein particle receptor biosynthetic process
          negative regulation of gluconeogenesis
          negative regulation of blood pressure
          positive regulation of blood pressure
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of fatty acid metabolic process
          positive regulation of glucose import
          negative regulation of hormone secretion
          negative regulation of smooth muscle cell proliferation
          negative regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of phagocytosis
          negative regulation of synaptic transmission
          brown fat cell differentiation
          protein homooligomerization
          response to glucocorticoid
          membrane depolarization
          membrane hyperpolarization
          protein heterotrimerization
          negative regulation of ERK1 and ERK2 cascade
          response to linoleic acid
          detection of oxidative stress
          cellular response to cAMP
          positive regulation of monocyte chemotactic protein-1 production
          cellular response to epinephrine stimulus
          protein localization to plasma membrane
          negative regulation of intracellular protein transport
          negative regulation of receptor binding
          negative regulation of DNA biosynthetic process
          positive regulation of glycogen (starch) synthase activity
          positive regulation of metanephric glomerular visceral epithelial cell development
          positive regulation of cAMP-dependent protein kinase activity
          positive regulation of renal albumin absorption
          negative regulation of platelet-derived growth factor receptor-alpha signaling pathway
          negative regulation of metanephric mesenchymal cell migration
          receptor binding
          hormone activity
          sialic acid binding
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P001089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169681 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001089 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001089 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P001089 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P393531 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755114 - -
          Affymetrix Rat Gene 1.0 ST Array 10755115 - -
          Affymetrix Rat Gene 1.0 ST Array 10755116 - -
          Affymetrix Rat Gene 1.1 ST Array 10755112 - -
          Rat Genome 230 2.0 Array 1381997_PM_at - -
          Rat Genome 230 2.0 Array 1381997_at - -
          Affymetrix HT Rat Focus Array 1381997_at - -
          Affymetrix HT Rat Focus Array 1381997_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100011949.rn.1 TC1100001880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011950.rn.1 TC1100001880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011944.rn.1 TC1100001880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011947.rn.1 TC1100001880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011943.rn.1 TC1100001880.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011942.rn.1 TC1100001880.rn.1 RTA TC
          Rat Expression Set 230 1381997_PM_at - -
          Rat Expression Set 230 1381997_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755112 - -
          Rat Genome U34 Set rc_AI105446_at - -
          Rat Genome U34 Set rc_AI176736_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22288946 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363454 - -
          RatRef-12 Expression BeadChip ILMN_56617 - -

          Back To Top
           

          Citations (1)

          Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
          Authors
          Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
          Journal
          J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 EDT 2009)
          Volume (Year)
          201 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline