Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595296_m1
          See other ASPG GE Assays ›
          Gene Symbol:
          Aspg
          Gene Name:
          asparaginase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 136682279 - 136701673 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595296_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aspg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144750.2 NP_653351.1 1 - 2 156 65
          GenBank mRNA AB009372.1 - 1 - 2 157 65
          BC098655.1 - 1 - 2 126 7373402 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001616.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aspg

          Gene Ontology Categories:

          Function(s) Process(es)

          asparagine metabolic process
          phospholipid metabolic process
          lipid catabolic process
          1-alkyl-2-acetylglycerophosphocholine esterase activity
          asparaginase activity
          lysophospholipase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23171 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148906 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148906 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148906 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P603700 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701934183 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89658 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45617 - -
          Agilent Rat cDNA Microarray (G4105A) 701934183 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10887456 - -
          Affymetrix Rat Gene 1.0 ST Array 10887450 - -
          Affymetrix Rat Gene 1.0 ST Array 10887448 - -
          Affymetrix Rat Gene 1.0 ST Array 10887447 - -
          Affymetrix Rat Gene 1.0 ST Array 10887451 - -
          Affymetrix Rat Gene 1.0 ST Array 10887457 - -
          Affymetrix Rat Gene 1.0 ST Array 10887452 - -
          Affymetrix Rat Gene 1.0 ST Array 10887458 - -
          Affymetrix Rat Gene 1.0 ST Array 10887455 - -
          Affymetrix Rat Gene 1.0 ST Array 10887454 - -
          Affymetrix Rat Gene 1.0 ST Array 10887453 - -
          Affymetrix Rat Gene 1.0 ST Array 10887460 - -
          Affymetrix Rat Gene 1.0 ST Array 10887449 - -
          Affymetrix Rat Gene 1.0 ST Array 10887446 - -
          Affymetrix Rat Gene 1.0 ST Array 10887459 - -
          Affymetrix Rat Gene 1.1 ST Array 10887445 - -
          Rat Genome 230 2.0 Array 1387959_PM_at - -
          Rat Genome 230 2.0 Array 1387959_at - -
          Affymetrix HT Rat Focus Array 1387959_at - -
          Affymetrix HT Rat Focus Array 1387959_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600011360.rn.1 TC0600001616.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600011361.rn.1 TC0600001616.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600011359.rn.1 TC0600001616.rn.1 RTA TC
          Rat Expression Set 230 1387959_at - -
          Rat Expression Set 230 1387959_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10887445 - -
          Rat Genome U34 Set AB009372_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21777361 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355386 - -
          RatRef-12 Expression BeadChip ILMN_62646 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline