Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595309_m1
          See other OAS1B GE Assays ›
          Gene Symbol:
          Oas1b
          Gene Name:
          2-5 oligoadenylate synthetase 1B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 41255762 - 41266430 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595309_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oas1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144752.2 NP_653353.2 5 - 6 1068 138
          GenBank mRNA AF068268.1 - 1080 138
          BC091121.1 - 5 - 6 1068 7383389 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001637.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oas1b

          Panther Classification:

          Molecular Function -

          nucleotidyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          response to virus
          negative regulation of viral genome replication
          innate immune response
          defense response to virus
          2'-5'-oligoadenylate synthetase activity
          double-stranded RNA binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731525 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025803 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332744 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702237155 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17134 - -
          Agilent Rat cDNA Microarray (G4105A) 702237155 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762249 - -
          Affymetrix Rat Gene 1.0 ST Array 10762251 - -
          Affymetrix Rat Gene 1.0 ST Array 10762252 - -
          Affymetrix Rat Gene 1.0 ST Array 10762253 - -
          Affymetrix Rat Gene 1.0 ST Array 10762248 - -
          Affymetrix Rat Gene 1.0 ST Array 10762250 - -
          Affymetrix Rat Gene 1.1 ST Array 10762247 - -
          Rat Genome 230 2.0 Array 1388056_PM_at - -
          Rat Genome 230 2.0 Array 1391754_PM_at - -
          Rat Genome 230 2.0 Array 1388056_at - -
          Rat Genome 230 2.0 Array 1391754_at - -
          Affymetrix HT Rat Focus Array 1388056_PM_at - -
          Affymetrix HT Rat Focus Array 1388056_at - -
          Affymetrix Rat Clariom S Assay PSR1200014014.rn.1 TC1200001637.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200014013.rn.1 TC1200001637.rn.1 RTA TC
          Rat Expression Set 230 1388056_at - -
          Rat Expression Set 230 1388056_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762247 - -
          Rat Genome U34 Set AF068268_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21122982 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362940 - -
          RatRef-12 Expression BeadChip ILMN_61316 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline