Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595311_m1
          See other CANT1 GE Assays ›
          Gene Symbol:
          Cant1
          Gene Name:
          calcium activated nucleotidase 1
          Gene Aliases:
          srapy
          Chromosome Location:
          Chr.10: 107432500 - 107445634 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595311_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cant1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144754.2 NP_653355.1 2 - 3 716 66
          XM_006247789.2 XP_006247851.1 2 - 3 1238 66
          XM_006247790.1 XP_006247852.1 2 - 3 786 66
          GenBank mRNA AJ312207.1 - 1 - 2 671 66
          BC097279.1 - 2 - 3 716 7443057 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004209.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cant1

          Panther Classification:

          Molecular Function -

          nucleotide phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          proteoglycan biosynthetic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          signal transducer activity
          calcium ion binding
          uridine-diphosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P255201 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P624726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670521 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33390 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P255201 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P624726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P255201 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P624726 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P255201 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P624726 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255201 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16891 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82482 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10749503 - -
          Affymetrix Rat Gene 1.0 ST Array 10749504 - -
          Affymetrix Rat Gene 1.0 ST Array 10749507 - -
          Affymetrix Rat Gene 1.0 ST Array 10749505 - -
          Affymetrix Rat Gene 1.0 ST Array 10749508 - -
          Affymetrix Rat Gene 1.1 ST Array 10749502 - -
          Rat Genome 230 2.0 Array 1389588_PM_at - -
          Rat Genome 230 2.0 Array 1396446_at - -
          Rat Genome 230 2.0 Array 1389588_at - -
          Rat Genome 230 2.0 Array 1396446_PM_at - -
          Affymetrix HT Rat Focus Array 1396446_PM_at - -
          Affymetrix HT Rat Focus Array 1389588_PM_at - -
          Affymetrix HT Rat Focus Array 1389588_at - -
          Affymetrix HT Rat Focus Array 1396446_at - -
          Affymetrix Rat Clariom S Assay PSR1000038499.rn.1 TC1000004209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000038497.rn.1 TC1000004209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000038504.rn.1 TC1000004209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000038500.rn.1 TC1000004209.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000038498.rn.1 TC1000004209.rn.1 RTA TC
          Rat Expression Set 230 1389588_at - -
          Rat Expression Set 230 1389588_PM_at - -
          Rat Expression Set 230 1396446_at - -
          Rat Expression Set 230 1396446_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10749502 - -
          Rat Genome U34 Set rc_AA946291_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21075712 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358720 - -
          RatRef-12 Expression BeadChip ILMN_55169 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline