Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595314_m1
          See other TRIB3 GE Assays ›
          Gene Symbol:
          Trib3
          Gene Name:
          tribbles pseudokinase 3
          Gene Aliases:
          NIPK
          Chromosome Location:
          Chr.3: 147814056 - 147819650 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595314_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trib3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144755.2 NP_653356.1 2 - 3 448 87
          GenBank mRNA AB020967.1 - 433 87
          BC091120.1 - 2 - 3 448 7326024 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004470.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trib3

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          protein phosphorylation
          negative regulation of protein kinase activity
          regulation of glucose transport
          programmed cell death
          positive regulation of protein binding
          cellular response to insulin stimulus
          response to endoplasmic reticulum stress
          regulation of MAP kinase activity
          negative regulation of fat cell differentiation
          negative regulation of fatty acid biosynthetic process
          negative regulation of transcription, DNA-templated
          positive regulation of ubiquitin-protein transferase activity
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          negative regulation of nucleic acid-templated transcription
          transcription corepressor activity
          protein kinase inhibitor activity
          ATP binding
          kinase activity
          enzyme binding
          protein kinase binding
          ubiquitin protein ligase binding
          ubiquitin-protein transferase regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581254 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117901 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117901 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117901 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P543774 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700696582 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95270 - -
          Agilent Rat cDNA Microarray (G4105A) 700696582 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10850781 - -
          Affymetrix Rat Gene 1.0 ST Array 10850778 - -
          Affymetrix Rat Gene 1.0 ST Array 10850777 - -
          Affymetrix Rat Gene 1.0 ST Array 10850779 - -
          Affymetrix Rat Gene 1.0 ST Array 10850780 - -
          Affymetrix Rat Gene 1.0 ST Array 10850782 - -
          Affymetrix Rat Gene 1.1 ST Array 10850775 - -
          Rat Genome 230 2.0 Array 1386321_s_at - -
          Rat Genome 230 2.0 Array 1370694_at - -
          Rat Genome 230 2.0 Array 1386321_PM_s_at - -
          Rat Genome 230 2.0 Array 1370695_PM_s_at - -
          Rat Genome 230 2.0 Array 1370694_PM_at - -
          Rat Genome 230 2.0 Array 1370695_s_at - -
          Affymetrix HT Rat Focus Array 1370695_s_at - -
          Affymetrix HT Rat Focus Array 1370695_PM_s_at - -
          Affymetrix HT Rat Focus Array 1370694_at - -
          Affymetrix HT Rat Focus Array 1386321_s_at - -
          Affymetrix HT Rat Focus Array 1370694_PM_at - -
          Affymetrix HT Rat Focus Array 1386321_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0300034337.rn.1 TC0300004470.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034335.rn.1 TC0300004470.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034336.rn.1 TC0300004470.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034334.rn.1 TC0300004470.rn.1 RTA TC
          Rat Expression Set 230 1370695_PM_s_at - -
          Rat Expression Set 230 1386321_s_at - -
          Rat Expression Set 230 1370694_PM_at - -
          Rat Expression Set 230 1370695_s_at - -
          Rat Expression Set 230 1386321_PM_s_at - -
          Rat Expression Set 230 1370694_at - -
          Affymetrix Rat Gene 1.0 ST Array 10850775 - -
          Rat Genome U34 Set rc_H31287_at - -
          Rat Genome U34 Set rc_H31287_g_at - -
          Rat Genome U34 Set rc_AI176901_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22163042 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349221 - -
          RatRef-12 Expression BeadChip ILMN_63345 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline