Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595407_m1
          See other ASRGL1 GE Assays ›
          Gene Symbol:
          Asrgl1
          Gene Name:
          asparaginase like 1
          Gene Aliases:
          Gliap, Hiob2
          Chromosome Location:
          Chr.1: 225309737 - 225329743 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595407_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Asrgl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145089.4 NP_659557.1 2 - 3 299 102
          GenBank mRNA AF329099.1 - 2 - 3 299 102
          AJ427914.2 - 313 102
          FQ212762.1 - 2 - 3 273 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007234.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Asrgl1

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          asparagine catabolic process via L-aspartate
          N4-(beta-N-acetylglucosaminyl)-L-asparaginase activity
          asparaginase activity
          beta-aspartyl-peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P002924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5651 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P002924 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P201077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44016 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728581 - -
          Affymetrix Rat Gene 1.0 ST Array 10728585 - -
          Affymetrix Rat Gene 1.0 ST Array 10728583 - -
          Affymetrix Rat Gene 1.0 ST Array 10728579 - -
          Affymetrix Rat Gene 1.0 ST Array 10728580 - -
          Affymetrix Rat Gene 1.0 ST Array 10728586 - -
          Affymetrix Rat Gene 1.0 ST Array 10728584 - -
          Affymetrix Rat Gene 1.0 ST Array 10728582 - -
          Affymetrix Rat Gene 1.1 ST Array 10728578 - -
          Rat Genome 230 2.0 Array 1387966_PM_at - -
          Rat Genome 230 2.0 Array 1375458_PM_at - -
          Rat Genome 230 2.0 Array 1374871_PM_at - -
          Rat Genome 230 2.0 Array 1374871_at - -
          Rat Genome 230 2.0 Array 1375458_at - -
          Rat Genome 230 2.0 Array 1387966_at - -
          Affymetrix HT Rat Focus Array 1387966_at - -
          Affymetrix HT Rat Focus Array 1387966_PM_at - -
          Affymetrix HT Rat Focus Array 1374871_PM_at - -
          Affymetrix HT Rat Focus Array 1374871_at - -
          Affymetrix HT Rat Focus Array 1375458_at - -
          Affymetrix HT Rat Focus Array 1375458_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100058379.rn.1 TC0100007234.rn.1 RTA TC
          Rat Expression Set 230 1374871_PM_at - -
          Rat Expression Set 230 1374871_at - -
          Rat Expression Set 230 1375458_at - -
          Rat Expression Set 230 1375458_PM_at - -
          Rat Expression Set 230 1387966_PM_at - -
          Rat Expression Set 230 1387966_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728578 - -
          Rat Genome U34 Set rc_AI232205_at - -
          Rat Genome U34 Set rc_AI233343_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21415489 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530180 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366911 - -
          RatRef-12 Expression BeadChip ILMN_54685 - -
          RatRef-12 Expression BeadChip ILMN_160558 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline