Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595448_m1
          See other SERPINA4 GE Assays ›
          Gene Symbol:
          Serpina4
          Gene Name:
          serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 127743883 - 127753047 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595448_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145097.2 NP_659565.2 3 - 4 961 122
          XM_008764741.1 XP_008762963.1 4 - 5 1189 122
          GenBank mRNA BC088111.1 - 3 - 4 961 7368874 122
          FQ209558.1 - 3 - 4 978 122
          FQ210636.1 - 3 - 4 981 122
          FQ218781.1 - 2 - 3 729 122
          U51017.1 - 2 - 3 922 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001398.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina4

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661665 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13410 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13410 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13410 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700608452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71629 - -
          Agilent Rat cDNA Microarray (G4105A) 700608452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886587 - -
          Affymetrix Rat Gene 1.0 ST Array 10886585 - -
          Affymetrix Rat Gene 1.0 ST Array 10886583 - -
          Affymetrix Rat Gene 1.0 ST Array 10886588 - -
          Affymetrix Rat Gene 1.0 ST Array 10886586 - -
          Affymetrix Rat Gene 1.0 ST Array 10886584 - -
          Affymetrix Rat Gene 1.1 ST Array 10886582 - -
          Rat Genome 230 2.0 Array 1370836_at - -
          Rat Genome 230 2.0 Array 1370836_PM_at - -
          Affymetrix HT Rat Focus Array 1370836_at - -
          Affymetrix HT Rat Focus Array 1370836_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600009767.rn.1 TC0600001398.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009766.rn.1 TC0600001398.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009768.rn.1 TC0600001398.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009765.rn.1 TC0600001398.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009769.rn.1 TC0600001398.rn.1 RTA TC
          Rat Expression Set 230 1370836_at - -
          Rat Expression Set 230 1370836_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886582 - -
          Rat Genome U34 Set rc_AA893552_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22386597 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352415 - -
          RatRef-12 Expression BeadChip ILMN_53616 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline