Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595467_m1
          See other USP15 GE Assays ›
          Gene Symbol:
          Usp15
          Gene Name:
          ubiquitin specific peptidase 15
          Gene Aliases:
          Ubp109
          Chromosome Location:
          Chr.7: 66595706 - 66687707 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595467_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usp15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145184.1 NP_660185.1 13 - 14 1796 100
          XM_006241431.2 XP_006241493.1 14 - 15 1914 100
          XM_008765376.1 XP_008763598.1 12 - 13 2226 100
          GenBank mRNA AF106657.1 - 13 - 14 1796 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000843.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Usp15

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          transforming growth factor beta receptor signaling pathway
          protein deubiquitination
          BMP signaling pathway
          monoubiquitinated protein deubiquitination
          histone H2B conserved C-terminal lysine deubiquitination
          proteasome-mediated ubiquitin-dependent protein catabolic process
          pathway-restricted SMAD protein phosphorylation
          regulation of proteasomal protein catabolic process
          cysteine-type endopeptidase activity
          thiol-dependent ubiquitin-specific protease activity
          transforming growth factor beta receptor binding
          identical protein binding
          SMAD binding
          ubiquitinated histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099332 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P189275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81012 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179473 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P225493 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P189275 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P189275 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P225493 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P225493 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P189275 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P225493 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700483028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63987 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46129 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11600 - -
          Agilent Rat cDNA Microarray (G4105A) 700483028 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10902848 - -
          Affymetrix Rat Gene 1.0 ST Array 10902856 - -
          Affymetrix Rat Gene 1.0 ST Array 10902849 - -
          Affymetrix Rat Gene 1.0 ST Array 10902858 - -
          Affymetrix Rat Gene 1.0 ST Array 10902844 - -
          Affymetrix Rat Gene 1.0 ST Array 10902855 - -
          Affymetrix Rat Gene 1.0 ST Array 10902846 - -
          Affymetrix Rat Gene 1.0 ST Array 10902852 - -
          Affymetrix Rat Gene 1.0 ST Array 10902854 - -
          Affymetrix Rat Gene 1.0 ST Array 10902851 - -
          Affymetrix Rat Gene 1.0 ST Array 10902847 - -
          Affymetrix Rat Gene 1.0 ST Array 10902845 - -
          Affymetrix Rat Gene 1.0 ST Array 10878220 - -
          Affymetrix Rat Gene 1.0 ST Array 10902850 - -
          Affymetrix Rat Gene 1.0 ST Array 10902853 - -
          Affymetrix Rat Gene 1.0 ST Array 10902857 - -
          Affymetrix Rat Gene 1.1 ST Array 10878219 - -
          Affymetrix Rat Gene 1.1 ST Array 10902843 - -
          Rat Genome 230 2.0 Array 1370460_at - -
          Rat Genome 230 2.0 Array 1370460_PM_at - -
          Affymetrix HT Rat Focus Array 1370460_PM_at - -
          Affymetrix HT Rat Focus Array 1370460_at - -
          Affymetrix Rat Clariom S Assay PSR0700006332.rn.1 TC0700000843.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700006333.rn.1 TC0700000843.rn.1 RTA TC
          Rat Expression Set 230 1370460_PM_at - -
          Rat Expression Set 230 1370460_at - -
          Affymetrix Rat Gene 1.0 ST Array 10902843 - -
          Rat Genome U34 Set rc_AA801313_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22034924 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370671 - -
          RatRef-12 Expression BeadChip ILMN_65177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline