Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00595525_m1
          See other CBY1 GE Assays ›
          Gene Symbol:
          Cby1
          Gene Name:
          chibby homolog 1 (Drosophila)
          Gene Aliases:
          Pgea1
          Chromosome Location:
          Chr.7: 120891928 - 120898318 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00595525_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cby1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145676.1 NP_663709.1 4 - 5 392 83
          XM_006241967.2 XP_006242029.1 5 - 6 509 83
          GenBank mRNA AF393211.1 - 4 - 5 392 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001455.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Rat Wnt Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cby1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization
          negative regulation of Wnt signaling pathway
          cilium assembly
          fat cell differentiation
          negative regulation of transcription, DNA-templated
          cardiac muscle cell differentiation
          negative regulation of canonical Wnt signaling pathway
          beta-catenin binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P139836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729965 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39699 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139836 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139836 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P139836 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897702 - -
          Affymetrix Rat Gene 1.0 ST Array 10897700 - -
          Affymetrix Rat Gene 1.0 ST Array 10897699 - -
          Affymetrix Rat Gene 1.0 ST Array 10897701 - -
          Affymetrix Rat Gene 1.1 ST Array 10897698 - -
          Rat Genome 230 2.0 Array 1397846_at - -
          Rat Genome 230 2.0 Array 1388859_PM_at - -
          Rat Genome 230 2.0 Array 1389654_PM_at - -
          Rat Genome 230 2.0 Array 1388859_at - -
          Rat Genome 230 2.0 Array 1389654_at - -
          Rat Genome 230 2.0 Array 1397846_PM_at - -
          Affymetrix HT Rat Focus Array 1397846_PM_at - -
          Affymetrix HT Rat Focus Array 1397846_at - -
          Affymetrix Rat Clariom S Assay PSR0700010916.rn.1 TC0700001455.rn.1 RTA TC
          Rat Expression Set 230 1388859_PM_at - -
          Rat Expression Set 230 1388859_at - -
          Rat Expression Set 230 1397846_PM_at - -
          Rat Expression Set 230 1397846_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897698 - -
          Rat Genome U34 Set rc_AA866257_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22337165 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350088 - -
          RatRef-12 Expression BeadChip ILMN_61731 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline